Skip to Content
Merck
All Photos(1)

Key Documents

96144

Sigma-Aldrich

Hyaluronic acid sodium salt from Streptococcus equi

mol wt 70,000-120,000

Synonym(s):

Sodium hyaluronate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

(Streptococcus equi)

Quality Level

form

powder

mol wt

70,000-120,000

color

white

solubility

water: ~5 g/L

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Na+].N([C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)C(=O)O)O)CO)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O[C@@H]3[C@H]([C@@H](O[C@@H]([C@H]3O)CO)O)NC(=O)C)C(=O)O)C(=O)C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hyaluronic acid, an anionic, nonsulfated glycosaminoglycan (GAG), may be used in research involving processes such as inflammation, cell signaling, cell migration, cell differentiation, tissue engineering and wound-healing. Hyaluronic acid may be used as a substrate to identify and characterized catabolizing enzymes such as hyaluronidase(s). Hyaluronic acid may be used in biomaterials research for the development of biological scaffolds for wound-healing applications and hyaluronic acid fillers in human skin.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shenglian Yao et al.
Journal of nanoscience and nanotechnology, 13(7), 4752-4758 (2013-08-02)
Nanofibers exist ubiquitously in natural extracellular matrix (ECM) of all kinds of human tissues forming hydrated interwoven network. Electrospinning nanotechnology has been proven to be a powerful technique to fabricate controllable nanofibers mimicking the natural ECM structures. Hyaluronic acid (HA)
[The dry eye].
Louis Tong et al.
Praxis, 102(13), 803-805 (2013-06-19)
Daniela Elena Costea et al.
Cancer research, 73(13), 3888-3901 (2013-04-20)
Heterogeneity of carcinoma-associated fibroblasts (CAF) has long been recognized, but the functional significance remains poorly understood. Here, we report the distinction of two CAF subtypes in oral squamous cell carcinoma (OSCC) that have differential tumor-promoting capability, one with a transcriptome
Incorporating facial rejuvenation procedures in the dental practice.
Warren Roberts
Today's FDA : official monthly journal of the Florida Dental Association, 25(2), 50-53 (2013-05-23)
M Gelardi et al.
European annals of allergy and clinical immunology, 45(3), 103-108 (2013-07-19)
To determine the effect of intranasal sodium hyaluronate on mucociliary clearance time following functional endoscopic sinus surgery in patients with nasal polyposis. Randomized, controlled, blinded study. Thirty-six patients with grade II nasal polyposis undergoing functional endoscopic sinus surgery received intranasal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service