Skip to Content
Merck
All Photos(1)

Key Documents

88190

Sigma-Aldrich

Tetrazolium Blue Chloride

≥90% (T), powder, suitable for microbiology

Synonym(s):

3,3′-(3,3′-Dimethoxy[1,1′-biphenyl]-4,4′-diyl)-bis(2,5-diphenyl-2H-tetrazolium) dichloride, 3,3′-Dianisole-4,4′-bis(3,5-diphenyltetrazolium chloride), BTC, Blue Tetrazolium chloride, TZ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H32Cl2N8O2
CAS Number:
Molecular Weight:
727.64
Beilstein:
3894077
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

Tetrazolium Blue Chloride, suitable for microbiology, ≥90% (T)

Quality Level

Assay

≥90% (T)

form

powder

mp

255 °C (dec.) (lit.)

solubility

methanol: 0.5 g/10 mL ( hot)

application(s)

microbiology

SMILES string

[Cl-].[Cl-].COc1cc(ccc1-[n+]2nc(nn2-c3ccccc3)-c4ccccc4)-c5ccc(c(OC)c5)-[n+]6nc(nn6-c7ccccc7)-c8ccccc8

InChI

1S/C40H32N8O2.2ClH/c1-49-37-27-31(23-25-35(37)47-43-39(29-15-7-3-8-16-29)41-45(47)33-19-11-5-12-20-33)32-24-26-36(38(28-32)50-2)48-44-40(30-17-9-4-10-18-30)42-46(48)34-21-13-6-14-22-34;;/h3-28H,1-2H3;2*1H/q+2;;/p-2

InChI key

MUUHXGOJWVMBDY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

Used in the assay of reducing sugars; Used in the preparation of DNA-cation complexes, immunoadsorbents for anti-DNA antibodies.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J.P. Robin et al.
Sciences Des Aliments, 1, 233-233 (1981)
Tetrazolium-based colorimetric assay for the detection of HIV replication inhibitors: revisited 20 years later.
Pannecouque C, Daelemans D, and De Clercq E
Nature Protocols, 3(3), 427-434 (2008)
V A DeBari et al.
Journal of immunological methods, 40(1), 89-94 (1981-01-01)
DNA can be precipitated as complexes with metal and organic cations. We have prepared these products using divalent metal cations (Cu2+, Mn2+, Ni2+, Zn2+, and Co2+) and ditetrazolium ions (blue tetrazolium and neotetrazolium). They are effective adsorbents, in vitro, for
Christophe Pannecouque et al.
Nature protocols, 3(3), 427-434 (2008-03-08)
Since its first description 20 years ago, the tetrazolium-based colorimetric (MTT) assay using MT-4 cells for the detection of anti-HIV compounds has been widely used. This method, which remains popular, provides more information than more recently developed methods and, therefore
Y Mizutani et al.
The Journal of urology, 160(2), 561-570 (1998-07-29)
The resistance of bladder cancer to anticancer chemotherapeutic drugs is a major problem. Several immunotherapeutic approaches have been developed to treat drug-resistant tumor cells. The Fas antigen (Fas)-Fas ligand pathway is involved in cytotoxic T lymphocyte and natural killer cell-mediated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service