Skip to Content
Merck
All Photos(1)

Key Documents

74680

Sigma-Aldrich

Octaethylene glycol monododecyl ether

BioXtra, ≥98.0% (GC)

Synonym(s):

C12E8, Dodecyl octaethylene glycol ether, Dodecyloctaglycol, Polyoxyethylene (8) lauryl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11(OCH2CH2)8OH
CAS Number:
Molecular Weight:
538.75
Beilstein:
1893466
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Quality Level

product line

BioXtra

Assay

≥98.0% (GC)

form

solid

mol wt

538.75 g/mol

CMC

0.11

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C28H58O9/c1-2-3-4-5-6-7-8-9-10-11-13-30-15-17-32-19-21-34-23-25-36-27-28-37-26-24-35-22-20-33-18-16-31-14-12-29/h29H,2-28H2,1H3

InChI key

YYELLDKEOUKVIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octaethylene glycol monododecyl ether is a nonionic detergent.

Application

Detergent used for the study of membrane proteins in a native-like state, e.g. ATPase. Used in a mixed micellar assay for lipoxygenase acting at neutral pH and for functional reconstitution of influenza virus envelopes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Relationship between activity and tetraprotomeric structure of ion-transporting ATPases].
Kazuhiro Abe et al.
Seikagaku. The Journal of Japanese Biochemical Society, 79(6), 527-534 (2007-08-01)
Cédric Montigny et al.
Biochemical and biophysical research communications, 391(1), 1067-1069 (2009-12-17)
Many membrane proteins become labile when they are solubilized by detergent. Here we show that the presence of high concentrations of glycyl betaine stabilizes one of these proteins, the sarcoplasmic reticulum Ca(2+)-ATPase (SERCA1a), solubilized with nonionic detergents like n-dodecyl beta-d-maltopyranoside
G C Troiano et al.
Biophysical journal, 75(2), 880-888 (1998-07-24)
The effects of a nonionic surfactant, octaethyleneglycol mono n-dodecyl ether (C12E8), on the electroporation of planar bilayer lipid membranes made of the synthetic lipid 1-pamitoyl 2-oleoyl phosphatidylcholine (POPC), was studied. High-amplitude ( approximately 100-450 mV) rectangular voltage pulses were used
C Indiveri et al.
The Biochemical journal, 333 ( Pt 2), 285-290 (1998-07-11)
The glutamine carrier from rat kidney mitochondria, solubilized in dodecyl octaoxyethylene ether (C12E8) and partly purified on hydroxyapatite, was identified and completely purified by Celite chromatography. On SDS/PAGE, the purified glutamine carrier consisted of a single protein band with an
M Bobrowska-Hägerstrand et al.
Biophysical journal, 77(6), 3356-3362 (1999-12-10)
Endovesicles induced in human erythrocytes by octaethyleneglycol dodecylether (C12E8) were studied by confocal laser scanning microscopy, using fluorescein isothiocyanate dextran as a nonspecific fluid marker. The endovesicles appeared to consist mainly of a ring-formed toroidal part joined with a central

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service