Skip to Content
Merck

Skip To

73641

Hyaluronic acid sodium salt from Streptococcus equi

mol wt 2,000,000-2,400,000

Synonym(s):

Sodium hyaluronate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Streptococcus equi)

Quality Level

form

powder

mol wt

2,000,000-2,400,000

color

white

solubility

water: ~5 g/L

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Na+].N([C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)C(=O)O)O)CO)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O[C@@H]3[C@H]([C@@H](O[C@@H]([C@H]3O)CO)O)NC(=O)C)C(=O)O)C(=O)C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
519676335780486
biological source

(Streptococcus equi)

biological source

(Streptococcus equi)

biological source

(Streptococcus equi)

biological source

(Streptococcus equi)

solubility

water: ~5 g/L

solubility

water: ~5 g/L

solubility

water: ~5 g/L

solubility

water: ~5 g/L

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

color

white

color

white

color

white

color

white


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Daniela Elena Costea et al.
Cancer research, 73(13), 3888-3901 (2013-04-20)
Heterogeneity of carcinoma-associated fibroblasts (CAF) has long been recognized, but the functional significance remains poorly understood. Here, we report the distinction of two CAF subtypes in oral squamous cell carcinoma (OSCC) that have differential tumor-promoting capability, one with a transcriptome
Shenglian Yao et al.
Journal of nanoscience and nanotechnology, 13(7), 4752-4758 (2013-08-02)
Nanofibers exist ubiquitously in natural extracellular matrix (ECM) of all kinds of human tissues forming hydrated interwoven network. Electrospinning nanotechnology has been proven to be a powerful technique to fabricate controllable nanofibers mimicking the natural ECM structures. Hyaluronic acid (HA)
M Gelardi et al.
European annals of allergy and clinical immunology, 45(3), 103-108 (2013-07-19)
To determine the effect of intranasal sodium hyaluronate on mucociliary clearance time following functional endoscopic sinus surgery in patients with nasal polyposis. Randomized, controlled, blinded study. Thirty-six patients with grade II nasal polyposis undergoing functional endoscopic sinus surgery received intranasal



Global Trade Item Number

SKUGTIN
73641-50MG04061838040558
73641-10MG04061838040541

Questions

Reviews

No rating value

Active Filters