Skip to Content
Merck
All Photos(1)

Key Documents

73341

Sigma-Aldrich

Echinenone

≥95.0% (HPLC), solid

Synonym(s):

β,β-Caroten-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H54O
CAS Number:
Molecular Weight:
550.86
Beilstein:
2030114
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Product Name

Echinenone, ≥95.0% (HPLC)

Quality Level

Assay

≥95.0% (HPLC)

form

solid

λ

in hexane (with 2% dichloromethane)

UV absorption

λ: 459 nm±5 nm Amax

storage temp.

−20°C

SMILES string

CC(/C=C/C1=C(C)C(CCC1(C)C)=O)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(\C=C\C2=C(C)CCCC2(C)C)C

InChI

1S/C40H54O/c1-30(18-13-20-32(3)23-25-36-34(5)22-15-28-39(36,7)8)16-11-12-17-31(2)19-14-21-33(4)24-26-37-35(6)38(41)27-29-40(37,9)10/h11-14,16-21,23-26H,15,22,27-29H2,1-10H3/b12-11+,18-13+,19-14+,25-23+,26-24+,30-16+,31-17+,32-20+,33-21+

InChI key

QXNWZXMBUKUYMD-QQGJMDNJSA-N

General description

Echinenone is a carotenoid with a conjugated carbonyl group. Echinenone (4-keto-β-carotene) is a monoketo compound, is an intermediate between β-carotene and canthaxanthin in animals. It is one of the major carotenoids of Anabaena sp., and also present in Micrococcus roseus. It is located in the thylakoid membrane of Anabaena. The conversion of β-carotene to echinenone is catalysed by CrtO, a β-carotene ketolase.

Application

Echinenone may be used as an external standard for the extraction of carotenoid from plasma samples. It may also been used as an internal standard, added to samples for extraction and quantification.

Biochem/physiol Actions

Echinenone binds to glocobacter rhodopsin and functions as a light harvesting antenna in Gloeobacter violaceous.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and identification of echinenone from Micrococcus roseus
Schwartzel EH and Cooney JJ
Journal of Bacteriology, 104(1), 272-274 (1970)
Reconstitution of gloeobacter rhodopsin with echinenone: role of the 4-keto group
Balashov SP, et al.
Biochemistry, 49(45), 9792-9799 (2010)
Echinenone vibrational properties: from solvents to the orange carotenoid protein
Kish E, et al.
Biochimica et Biophysica Acta - Bioenergetics, 1847(10), 1044-1054 (2015)
Immune status is more affected by age than by carotenoid depletion-repletion in healthy human subjects
Farges MC, et al.
The British Journal of Nutrition, 108(11), 2054-2065 (2012)
Faiza Hammaz et al.
Food chemistry, 338, 128004-128004 (2020-09-21)
The aim was to enhance provitamin A carotenoid (proVA CAR) concentrations and bioaccessibility in carrots by manipulating post-harvest factors. To that end, we assessed the effects of Ultraviolet-C light, pulsed light, storage temperature, and storage duration. We also measured CAR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service