Skip to Content
Merck
All Photos(1)

Key Documents

69553

Sigma-Aldrich

α-Cryptoxanthin

≥90.0% (HPLC)

Synonym(s):

α-Caroten-3-ol, (3R,6′R)-β,ε-Caroten-3-ol, Physoxanthin, Zeinoxanthin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56O
CAS Number:
Molecular Weight:
552.87
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥90.0% (HPLC)

form

solid

λ

in hexane (with 2% dichloromethane)

UV absorption

λ: 450 nm±10 nm Amax

storage temp.

−20°C

SMILES string

CC(/C=C/C1=C(C)C[C@@H](O)CC1(C)C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(\C=C\[C@@H]2C(C)(C)CCC=C2C)C

InChI

1S/C40H56O/c1-30(18-13-20-32(3)23-25-37-34(5)22-15-27-39(37,7)8)16-11-12-17-31(2)19-14-21-33(4)24-26-38-35(6)28-36(41)29-40(38,9)10/h11-14,16-26,36-37,41H,15,27-29H2,1-10H3/b12-11+,18-13+,19-14+,25-23+,26-24+,30-16+,31-17+,32-20+,33-21+/t36-,37+/m1/s1

InChI key

NBZANZVJRKXVBH-NHWXEJKLSA-N

General description

α-Cryptoxanthin is a derivative of asymmetric α-carotene (β,ε-carotene) and belongs to the carotenoid family. It is one of the major xanthophylls present in the human brain. α-Cryptoxanthin is found in carrot leaves and autumn olive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jörg Schlatterer et al.
Journal of agricultural and food chemistry, 53(16), 6355-6361 (2005-08-04)
Citrus fruits contain a wide range of bioactive compounds. Their carotenoid fraction is inter alia dominated by structural cryptoxanthin isomers as beta-cryptoxanthin and zeinoxanthin. Both xanthophylls were identified in saponified citrus fruit extracts by comparison to reference compounds extracted from
Fruit of Autumn Olive: A Rich Source of Lycopene
Fordham I M, et al.
Hortscience: a Publication of the American Society For Horticultural Science Hortscience, 36(3), 1136-1137 (2001)
Analysis of ?-Cryptoxanthin, ?-Cryptoxanthin, ? -Carotene, and ?-Carotene of Pandanus Conoideus Oil by High-performance Liquid Chromatography (HPLC)
Sarungallo Z L, et al.
Procedia Food Science, 3, 231-243 (2015)
Jörg Schlatterer et al.
The British journal of nutrition, 96(2), 371-376 (2006-08-23)
Xanthophylls have attracted a lot of interest since their health benefits were documented. Unfortunately, studying their intestinal absorption is often affected by high baseline levels present in the fasting plasma. As alpha-cryptoxanthin is rarely found in the traditional European diet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service