Skip to Content
Merck
All Photos(1)

Key Documents

68530

Sigma-Aldrich

Pinobanksin

≥95% (HPLC)

Synonym(s):

(2R,3R)-3,5,7-Trihydroxy-2-phenyl-2,3-dihydro-4H-chromen-4-one, (2R,3R)-3,5,7-Trihydroxy-2-phenyl-chroman-4-one, (2R,3R)-3,5,7-Trihydroxyflavanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
Beilstein:
91238
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

O[C@@H]1[C@H](Oc2cc(O)cc(O)c2C1=O)c3ccccc3

InChI

1S/C15H12O5/c16-9-6-10(17)12-11(7-9)20-15(14(19)13(12)18)8-4-2-1-3-5-8/h1-7,14-17,19H/t14-,15+/m0/s1

InChI key

SUYJZKRQHBQNCA-LSDHHAIUSA-N

General description

Pinobanksin is a flavonoid that is widely available in honey and different propolis.

Application

Pinobanksin has been used as a reference standard to identify and measure its presence in eleven different acacia honey samples using high-performance liquid chromatography with photo-diode array detection (HPLC-PDA).

Biochem/physiol Actions

Pinobanksin is a potent antioxidant. It displays several anti-cancer properties as an apoptosis inducer, anti-proliferative and antiangiogenic agent in B-cell lymphoma (M12.C3.F6) cells and human umbilical vein endothelial cells, respectively. Pinobanksin also exerts antimicrobial and antifungal properties.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan-Zhen Zheng et al.
Food chemistry, 240, 323-329 (2017-09-28)
The effects of the ester group and solvent on the structure and antioxidant activity of pinobanksin were carried out using DFT calculation. First, the properties of the intramolecular hydrogen-bonds in the investigated compounds were studied. Second, the antioxidant capacities of
Antiangiogenic effect of pinobanksin on human umbilical vein endothelial cells
Bang, Ho-Jeong and Ahn, Mok-Ryeon
Journal of functional foods, 79, 104408-104408 (2021)
Xiaolan Xu et al.
Molecules (Basel, Switzerland), 24(19) (2019-10-09)
The chemical compositions of ethanol extracts of propolis from China (EEP-C) and the United States (EEP-A) and their antifungal activity against Penicillium notatum were determined. The result showed that a total of 49 compounds were detected by UPLC-Q-TOF-MS, 30 of
Potential antimicrobial activity of honey phenolic compounds against Gram positive and Gram negative bacteria
Leyva-Jimenez F J, et al.,
Lebensm. Wiss. Technol., 101, 236-245 (2019)
L Krizková et al.
Mutation research, 416(1-2), 85-92 (1998-09-03)
The antimutagenicity of 14 naturally occurring flavonoids (20 mumol/l) on ofloxacin (43 mumol/l and 86 mumol/l)-induced bleaching (mutagenicity) was studied in Euglena gracilis. The flavonoids chrysin, techtochrysin, chrysin-5-methylether galangin, galangin-5-methylether, pinocembrin and pinobanksin possess considerable antimutagenic properties against ofloxacin-induced bleaching

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service