Skip to Content
Merck
All Photos(2)

Key Documents

95082

Supelco

Chrysin

analytical standard

Synonym(s):

5,7-Dihydroxyflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O4
CAS Number:
Molecular Weight:
254.24
Beilstein:
233276
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

284-286 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Oc1cc(O)c2C(=O)C=C(Oc2c1)c3ccccc3

InChI

1S/C15H10O4/c16-10-6-11(17)15-12(18)8-13(19-14(15)7-10)9-4-2-1-3-5-9/h1-8,16-17H

InChI key

RTIXKCRFFJGDFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chrysin is a naturally occurring mono-flavonoid , which is present in plant cells involved in photosynthesis. It may possess a broad spectrum of pharmacological properties like antioxidant, anti-viral, anti-inflammatory, anxiolytic and vasodilatory effects.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Principles of Orthomolecularism (2004)
Possible anxiolytic effects of chrysin, a central benzodiazepine receptor ligand isolated from Passiflora coerulea
Wolfman C, et al.
Pharmacology, Biochemistry, and Behavior, 47(1), 1-4 (1994)
G A R Oliveira et al.
Journal of toxicology and environmental health. Part A, 75(16-17), 1000-1011 (2012-08-03)
Chrysin is one of the natural flavonoids present in plants, and large amounts are present in honey and propolis. In addition to anticancer, antioxidation, and anti-inflammatory activities, chrysin has also been reported to be an inhibitor of aromatase, an enzyme
Rehan Khan et al.
The British journal of nutrition, 108(9), 1574-1585 (2012-02-09)
Cisplatin (cis-diamminedichloroplatinum (II) (CDDP)) is a commonly used chemotherapeutic drug for the treatment of numerous forms of cancer, but it has pronounced adverse effects, namely nephrotoxicity, ototoxicity, neurotoxicity, hepatotoxicity, diarrhoea and nausea. CDDP-induced emesis and diarrhoea are also marked toxicities
Alexandrine During et al.
Biochemical pharmacology, 86(12), 1739-1746 (2013-10-19)
The aim of this study was to investigate whether methoxylated flavones versus their unmethylated analogs can modulate the intestinal inflammatory response. Flavone effects were assessed on soluble pro-inflammatory mediator (IL-8, IL-6, macrophage chemoattractant protein-1 (MCP-1), and cyclooxygenase-2 (COX-2)-derived PGE2) production

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service