Skip to Content
Merck
All Photos(1)

Key Documents

51198

Sigma-Aldrich

Gum arabic from acacia tree

spray dried

Synonym(s):

Acacia gum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

plant (acacia)

Quality Level

form

powder

quality

spray dried

ign. residue

≤4%

loss

≤10% loss on drying

color

white to faint beige

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Given orally, acacia gum ameliorates the severity of chronic renal failure (CRF) in humans and rats. Acacia gum is used as a surfactant in the development of hydrogels for tissue engineering applications. Acacia gum polymer is used to preserve microorganisms during sampling and storage.

Other Notes

Believed to be a branched polymer of galactose, rhamnose, arabinose, and glucuronic acid as the calcium, magnesium, and potassium salts with a mol. wt. of approx. 250,000.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of Gum Arabic in rats with adenine-induced chronic renal failure.
Ali BH, Al-Salam S, Al Husseni I, et al.
Exp. Biol. Med, 235, 373-382 (2010)
Effect of Acacia gum on blood pressure in rats with adenine-induced chronic renal failure.
Ali BH, Ziada A, Al Husseni I, et al.
Phytochemistry, 18, 1176-1180 (2011)
Iryna Sorokulova et al.
Journal of microbiological methods, 88(1), 140-146 (2011-11-19)
Stability of microbial cultures during sampling and storage is a vital issue in various fields of medicine, biotechnology, food science, and forensics. We have developed a unique bacterial preservation process involving a non-toxic, water-soluble acacia gum polymer that eliminates the
Chengdong Ji et al.
Biomaterials, 32(36), 9719-9729 (2011-09-20)
The aim of this study was to develop a process to create highly porous three-dimensional (3D) chitosan hydrogels suitable for tissue engineering applications. Chitosan was crosslinked by glutaraldehyde (0.5 vol %) under high pressure CO(2) at 60 bar and 4
Krista M Sumby et al.
Applied and environmental microbiology, 75(21), 6729-6735 (2009-09-08)
We report the cloning and characterization of EstB28, the first esterase to be so characterized from the wine-associated lactic acid bacterium, Oenococcus oeni. The published sequence for O. oeni strain PSU-1 was used to identify putative esterase genes and design

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service