44001
N-Acetyl-D-glucosamine 6-sulfate sodium salt
≥98.0% (TLC)
Synonym(s):
GlcNAc-6S
About This Item
Recommended Products
biological source
synthetic
description
Mixed anomers
Assay
≥98.0% (TLC)
form
solid
optical activity
[α]20/D 32.0±2.0, c = 0.1% (w/v) in water
technique(s)
thin layer chromatography (TLC): suitable
color
colorless
storage temp.
−20°C
SMILES string
[Na+].CC(=O)N[C@H]1C(O)O[C@H](COS([O-])(=O)=O)[C@@H](O)[C@@H]1O
InChI
1S/C8H15NO9S.Na/c1-3(10)9-5-7(12)6(11)4(18-8(5)13)2-17-19(14,15)16;/h4-8,11-13H,2H2,1H3,(H,9,10)(H,14,15,16);/q;+1/p-1/t4-,5-,6-,7-,8?;/m1./s1
InChI key
CBUJZKTVEFVBBG-FROKLYQUSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Packaging
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service