Skip to Content
Merck

Skip To

41009

Polyethylene glycol solution

12% PEG 8000 – Tris pH 8; 100 mM

Synonym(s):

Crystallization solution 34/Kit-No 02528, Polyethylene glycol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
51171641
eCl@ss:
32161000
NACRES:
NA.28
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

PEG 8000 12% – Tris pH 8; 100 mM solution,

form

liquid

Quality Segment

pH

8.0±0.2 ( in neat)

density

1.02 g/mL at 20 °C

storage temp.

2-8°C

Application


  • Polymorphism of microcrystalline urate oxidase from Aspergillus flavus.: This study explores the polymorphic behaviors of urate oxidase crystals under various chemical environments, highlighting modifications in crystal morphology and stability in the presence of additives like polyethylene glycol (PEG) and pH buffers (Collings et al., 2010).

  • Crystallization and preliminary X-ray analysis of a recombinant Fab fragment in complex with 17beta-oestradiol.: Details the crystallization process of a Fab fragment bound with 17beta-oestradiol, utilizing a crystallization medium enriched with PEG 8000 and Tris buffer, analyzing its impact on crystal quality and data collection (Lamminmaki and Kankare, 2000).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
11130
form

liquid

form

-

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

density

1.02 g/mL at 20 °C

density

1.03 g/mL at 20 °C

pH

8.0±0.2 ( in neat)

pH

8.0±0.2 ( in neat)


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ryan McCormack et al.
Journal of immunology (Baltimore, Md. : 1950), 204(8), 2242-2256 (2020-03-13)
Type I IFNs play a complex role in determining the fate of microbial pathogens and may also be deleterious to the host during bacterial and viral infections. Upon ligand binding, a receptor proximal complex consisting of IFN-α and -β receptors



Global Trade Item Number

SKUGTIN
41009-100ML-F04061837797729
41009-10ML-KC-F04061824679632

Questions

Reviews

No rating value

Active Filters