40530
Poly(ethylene glycol) octyl ether
Synonym(s):
C8En (n = 2 to 9), Polyethylene glycol, n-Octylpolyoxyethylene, Octyl-POE, Polyethylene glycol monooctyl ether, Rosenbusch-Tenside
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)6CH2(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25
Recommended Products
form
liquid
refractive index
n20/D 1.452
SMILES string
O(CCO)CCCCCCCC
InChI
1S/C10H22O2/c1-2-3-4-5-6-7-9-12-10-8-11/h11H,2-10H2,1H3
InChI key
ZQCIMPBZCZUDJM-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Stéphane Abel et al.
Biochimica et biophysica acta. Biomembranes, 1863(2), 183504-183504 (2020-11-07)
Successful crystallization of membrane proteins in detergent micelles depends on key factors such as conformational stability of the protein in micellar assemblies, the protein-detergent complex (PDC) monodispersity and favorable protein crystal contacts by suitable shielding of the protein hydrophobic surface
Jesper E Mogensen et al.
Protein science : a publication of the Protein Society, 14(11), 2814-2827 (2005-10-04)
The adhesin involved in diffuse adherence (AIDA) is an autotransporter protein that confers the diffuse adherence phenotype to certain diarrheagenic Escherichia coli strains. It consists of a 49 amino acid signal peptide, a 797 amino acid passenger domain, and a
C Ostermeier et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(20), 10547-10553 (1997-10-06)
The aa3 type cytochrome c oxidase consisting of the core subunits I and II only was isolated from the soil bacterium Paracoccus denitrificans and crystallized as complex with a monoclonal antibody Fv fragment. Crystals could be grown in the presence
Purification and partial characterization of the OmpA family of proteins of Pasteurella haemolytica.
P J Mahasreshti et al.
Infection and immunity, 65(1), 211-218 (1997-01-01)
This study was conducted to partially characterize and identify the purity of two major outer membrane proteins (OMPs) (with molecular weights of 32,000 and 35,000 [32K and 35K, respectively]) of Pasteurella haemolytica. The 35K and 32K major OMPs, designated Pasteurella
Hester Emilie Hasper et al.
Biochemistry, 43(36), 11567-11575 (2004-09-08)
The peptide antibiotic nisin was the first reported example of an antibiotic that kills bacteria via targeted pore formation. The specific target of nisin is Lipid II, an essential intermediate in the bacterial cell-wall synthesis. High-affinity binding of the antibiotic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service