Skip to Content
Merck
All Photos(1)

Documents

40236

Sigma-Aldrich

N,N-Dimethyldodecylamine N-oxide solution

~30% in H2O

Synonym(s):

DDAO, LDAO, Lauryldimethylamine N-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H31NO
CAS Number:
Molecular Weight:
229.40
Beilstein:
1769927
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

form

liquid

mol wt

229.40 g/mol

concentration

~30% in H2O

technique(s)

protein quantification: suitable

refractive index

n20/D 1.378

density

0.982 g/mL at 20 °C

SMILES string

CCCCCCCCCCCC[N+](C)([O-])C

InChI

1S/C14H31NO/c1-4-5-6-7-8-9-10-11-12-13-14-15(2,3)16/h4-14H2,1-3H3

InChI key

SYELZBGXAIXKHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethyldodecylamine N-oxide (DDAO), an amphoteric surfactant, is nonionic at high pH. The surfactant headgroup of amphoteric surfactants may carry positive and negative charges as the net charge is pH sensitive.

Application

N,N-Dimethyldodecylamine N-oxide solution has been used:
  • to examine the individual components of solvable for the extraction of few-layer graphene (FLG)and graphene oxide (GO) from complex organic matrices
  • as a component of wash buffer, elution buffer, and anion loading buffer for the purification of the double mutant reaction centers (DM RCs)
  • in the extraction of bilirubin and biliverdin by high-performance liquid chromatography (HPLC)

Biochem/physiol Actions

N,N-Dimethyldodecylamine N-oxide (DDAO) is preferred for protein studies. It is also frequently utilized for the extraction of membrane proteins.

Features and Benefits

  • Suitable for protein quantification

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Jun et al.
STAR protocols, 3(1), 101044-101044 (2022-01-04)
The formation of defined surfaces consisting of photosynthetic reaction centers (RCs) in biohybrid solar cells is challenging. Here, we start with the production of engineered RCs for oriented binding. RCs are deposited onto gold electrodes, and 6-mercapto-1-hexanol (MCH) is used
Karl B McCann et al.
Biologicals : journal of the International Association of Biological Standardization, 37(1), 32-36 (2008-10-25)
Human serum albumin is a well tolerated therapeutic for the treatment of hypovolemia. Despite all commercial human albumin preparations being derived from plasma, these products can have a highly variable colour. Albumin samples derived from ethanol precipitation and chromatographic fractionation
David S White et al.
eLife, 9 (2020-04-09)
Single-molecule approaches provide enormous insight into the dynamics of biomolecules, but adequately sampling distributions of states and events often requires extensive sampling. Although emerging experimental techniques can generate such large datasets, existing analysis tools are not suitable to process the
Kayla D Thompson et al.
Langmuir : the ACS journal of surfaces and colloids, 38(13), 4090-4101 (2022-03-25)
Understanding the interactions between surfactants and proteins is important for the formulation of consumer products as surfactant binding can alter protein activity and stability. Additionally, the structure of the protein-surfactant complex can influence surface activity, which is important for emulsion
Thomas Kupke et al.
Communications biology, 3(1), 73-73 (2020-02-16)
Transmembrane signaling proteins play a crucial role in the transduction of information across cell membranes. One function of regulated intramembrane proteolysis (RIP) is the release of signaling factors from transmembrane proteins. To study the role of transmembrane domains (TMDs) in modulating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service