Skip to Content
Merck
All Photos(1)

Documents

1.09033

Sigma-Aldrich

Schiff′s reagent

for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.41

Quality Level

form

liquid

IVD

for in vitro diagnostic use

pH

2.1-2.5 (25 °C in H2O)

density

1.01 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Schiff‘s reagent -for microscopy for the periodic acid Schiff, or PAS reaction, is a ready-to-use solution used for the detection of mucopolysaccharides, glycogen, muco- and glycoproteins, glyco-, phospholipids, basal membranes and collagen. The PAS (periodic acid Schiff) reaction is one of the most frequently used chemical methods for the detection of aldehyde and mucosubstances in histology. In the PAS reaction, the histological specimen material is first treated with periodic acid, resulting in the oxidation of the 1,2-glycols into aldehyde groups. The addition of Schiff′s reagent (fuchsin-sulfuric acid) in the second step causes the aldehydes to react to form a brilliant red color. In the end result, the PAS reaction yields a specific color reaction with unsubstituted polysaccharides, neutral mucopolysaccharides, muco- and glycoproteins, and glyco- and phospholipids. The PAS reaction can be further combined with the Alcian blue staining method to detect mucosubstances (glycosaminoglycans). 500 ml are sufficient for approx. 2500 - 3000 stainings. It is registered as IVD and CE certified product and can be used for clinical diagnostics. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

For microscopy. Ready-to-use staining solution for human-medical cell diagnosis.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorival Mendes Rodrigues-Junior et al.
Investigational new drugs, 38(1), 70-78 (2019-05-19)
Purpose Among alkaloids, abundant secondary metabolites in plants, aporphines constitute a class of compounds with interesting biological activities, including anticancer effects. The present study evaluated the anticancer activities of 14 substances, including four aporphine derivatives acquired through the biomonitoring of
D Piacentini et al.
Plant physiology and biochemistry : PPB, 151, 729-742 (2020-05-01)
Nitric oxide (NO) has signalling roles in plant stress responses. Cadmium (Cd) and arsenic (As) soil pollutants alter plant development, mainly the root-system, by increasing NO-content, triggering reactive oxygen species (ROS), and forming peroxynitrite by NO-reaction with the superoxide anion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service