Skip to Content
Merck
All Photos(1)

Documents

382M-1

Sigma-Aldrich

SOX-11 (MRQ-58) Mouse Monoclonal Antibody

clone MRQ-58, unconjugated, Cell Marque

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-58, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (382M-14)
vial of 0.5 mL concentrate (382M-15)
bottle of 1.0 mL predilute (382M-17)
vial of 1.0 mL concentrate (382M-16)
bottle of 7.0 mL predilute (382M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1

control

mantle cell lymphoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... SOX11(6664)

General description

SOX-11 which is a member of the SOX (SRY-related HMG-box) family is a transcription factor normally expressed in the developing human central nervous system and plays a role in embryonic cell determination. citation Studies show that SOX-11 can be used as a marker for mantle cell lymphoma (MCL).

Quality


IVD

IVD

IVD

RUO

Linkage

SOX-11 Positive Control Slides, Product No. 382S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mantle cell lymphoma: from pathology and molecular pathogenesis to new therapeutic perspectives.
Itziar Salaverria et al.
Haematologica, 91(1), 11-16 (2006-01-26)
Tiemo Katzenberger et al.
British journal of haematology, 142(4), 538-550 (2008-06-20)
Mantle cell lymphoma (MCL) is an aggressive lymphoid tumour characterized by the translocation t(11;14)(q13;q32) and a poor clinical outcome (median survival: 3-4 years). Recent studies revealed that increased proliferation of the tumour cells and certain chromosomal aberrations, such as deletions
Bernd Weigle et al.
Oncology reports, 13(1), 139-144 (2004-12-08)
Malignant glioma comprises the majority of primary human brain tumors with 16,800 new cases reported each year in the USA. Its prognosis remains dismal despite numerous attempts to improve conventional therapeutic modalities. Therefore, much effort is devoted to the exploration
Kai Fu et al.
Blood, 106(13), 4315-4321 (2005-08-27)
Cyclin D1 overexpression is believed to be essential in the pathogenesis of mantle cell lymphoma (MCL). Hence, the existence of cyclin D1-negative MCL has been controversial and difficult to substantiate. Our previous gene expression profiling study identified several cases that
Ching-Jung Lee et al.
Journal of neuro-oncology, 57(3), 201-214 (2002-07-20)
Primitive neuroectodermal tumors (PNETs) are composed of immature neuronal precursor cells and sometimes more mature neuronal cell types. Medulloblastomas, occuring in the cerebellum, represent the most common PNET and are broadly classified into two subgroups: classical and desmoplastic. Desmoplastic medulloblastomas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service