Skip to Content
Merck
All Photos(1)

Documents

367M-1

Sigma-Aldrich

IgG4 (MRQ-44) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-44, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (367M-14)
vial of 0.5 mL concentrate (367M-15)
bottle of 1.0 mL predilute (367M-17)
vial of 1.0 mL concentrate (367M-16)
bottle of 7.0 mL predilute (367M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

The igG4-related sclerosing disease has been recognized as a systemic disease entity characterized by an elevated serum IgG4 level, sclerosing fibrosis, and diffuse lymphoplasmacytic infiltration with the presence of many IgG4-positive plasma cells. Clinical manifestations are apparent in the pancreas, bile duct, gall bladder, lacrimal gland, salivary gland, retroperitoneum, kidney, lung, breast, thyroid, and prostate. Immunohistochemical analyses in the case of IgG4-related sclerosing disease not only exhibit significantly more than normal IgG4-positive plasma cells in affected tissues.

Quality


IVD

IVD

IVD

RUO

Linkage

IgG4 Positive Control Slides, Product No. 367S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noriyuki Sakata et al.
The American journal of surgical pathology, 32(4), 553-559 (2008-02-28)
Inflammatory abdominal aortic aneurysm (IAA) is associated with autoimmune disease. However, the precise mechanism of IAA remains unclear. There is increasing evidence that IgG4 is involved in the autoimmune mechanism of various idiopathic sclerosing lesions, including sclerosing pancreatitis and retroperitoneal
Sudhir Dhobale et al.
Journal of clinical rheumatology : practical reports on rheumatic & musculoskeletal diseases, 15(7), 354-357 (2009-12-17)
Great attention has been drawn toward the recently defined IgG4 related sclerosing disease, an entity incorporating autoimmune pancreatitis and other organ involvements, hypergammaglobulinemia and deposition of IgG4 in affected tissues. We demonstrate an index case of this disease with multiple
Yasuharu Sato et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 22(4), 589-599 (2009-03-10)
IgG4-related disease sometimes involves regional and/or systemic lymph nodes, and often clinically and/or histologically mimics multicentric Castleman's disease or malignant lymphoma. In this study, we examined clinical and pathologic findings of nine patients with systemic IgG4-related lymphadenopathy. None of these
Wah Cheuk et al.
The American journal of surgical pathology, 33(7), 1058-1064 (2009-04-23)
Immunoglobulin G (IgG)4-related sclerosing disease is a recently described syndrome characterized by mass-forming lesions in various organs due to dense lymphoplasmacytic infiltrates and stromal sclerosis, elevated serum IgG4 titer, increased tissue IgG4 plasma cells, and favorable clinical outcome. We describe
Terumi Kamisawa et al.
World journal of gastroenterology, 15(19), 2357-2360 (2009-05-20)
To clarify the characteristic features of biliary lesions in patients with autoimmune pancreatitis (AIP) and compare them with those of primary sclerosing cholangitis (PSC). The clinicopathological characteristics of 34 patients with sclerosing cholangitis (SC) associated with AIP were compared with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service