Skip to Content
Merck
All Photos(1)

Documents

31404

Sigma-Aldrich

Dextran sulfate sodium salt from Leuconostoc spp.

Mr 5,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

biological source

(Leuconostoc spp.)

form

powder

mol wt

Mr 5,000

composition

Sulfur Content, 16.0-20.0%

loss

≤10% loss on drying

useful pH range

6.0-8.0 (10 g/L)

solubility

H2O: 0.1 g/mL, clear, colorless to slightly yellow
water: 100 g/L at 25 °C (77 °F )
H2O: freely soluble 50 mg/mL, clear

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Dextran sulfate sodium salt from Leuconostoc sp. is a polyanion used as a test reagent for testing its capability to rupture the extracellular enveloped virus (EEV) outer membrane. It has also been used for assessing the impact of TLR4 monoclonal antibody on the growth of dextran-sulfate-sodium-(DSS)-induced colitis.

Biochem/physiol Actions

Dextran is a mixture of glucose polymers of varying size and geometry.Dextran in the form of dextran sulphate sodium acts as an anticoagulant. Dextran interferes with blood clot formation by acting on blood coagulation factors and the function of platelets. It prolongs the time required for formation of the clot and decreases the maximum clot strength. The disturbance of homeostasis by dextran may cause increased bleeding, colitis and kidney failure.

Preparation Note

prepared from dextran with Mr ~5000

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Liu et al.
Journal of gastroenterology and hepatology, 25(1), 209-214 (2009-11-26)
Ulcerative colitis (UC) refers to a kind of inflammatory bowel disease, of which the accurate pathogenesis is not yet well understood. Recently, the toll-like receptor 4 (TLR4) and the TLR4 signaling pathway have been proved as playing an important role
Simon Finfer et al.
The New England journal of medicine, 350(22), 2247-2256 (2004-05-28)
It remains uncertain whether the choice of resuscitation fluid for patients in intensive care units (ICUs) affects survival. We conducted a multicenter, randomized, double-blind trial to compare the effect of fluid resuscitation with albumin or saline on mortality in a
Kim L Roberts et al.
The Journal of general virology, 90(Pt 7), 1582-1591 (2009-03-07)
The extracellular enveloped virus (EEV) form of vaccinia virus (VACV) is surrounded by two lipid envelopes. This presents a topological problem for virus entry into cells, because a classical fusion event would only release a virion surrounded by a single
Lisa Nebelin Hvidt et al.
Danish medical journal, 59(11), A4531-A4531 (2012-11-23)
Synthetic colloids are frequently used in fluid resuscitation of septic patients. Despite this, little is known about the potential side effects including the risk of renal failure and bleeding. As practice has changed, we performed a before-and-after study of fluid
Thomas Böttcher et al.
Applied and environmental microbiology, 84(13) (2018-04-22)
Only a small fraction of bacteria can autonomously initiate growth on agar plates. Nongrowing bacteria typically enter a metabolically inactive dormant state and require specific chemical trigger factors or signals to exit this state and to resume growth. Micrococcus luteus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service