Skip to Content
Merck
All Photos(1)

Key Documents

30891

Sigma-Aldrich

Biliverdin hydrochloride

≥97.0% (TLC)

Synonym(s):

Biliverdin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H34N4O6 · HCl
CAS Number:
Molecular Weight:
619.11
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97.0% (TLC)

form

powder or crystals

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

Cl.CC1=C(C=C)C(\NC1=O)=C\c2[nH]c(/C=C3\N=C(\C=C4/NC(=O)C(C=C)=C4C)C(C)=C3CCC(O)=O)c(CCC(O)=O)c2C

InChI

1S/C33H34N4O6.ClH/c1-7-20-19(6)32(42)37-27(20)14-25-18(5)23(10-12-31(40)41)29(35-25)15-28-22(9-11-30(38)39)17(4)24(34-28)13-26-16(3)21(8-2)33(43)36-26;/h7-8,13-15,35H,1-2,9-12H2,3-6H3,(H,36,43)(H,37,42)(H,38,39)(H,40,41);1H/b26-13-,27-14-,28-15-;

InChI key

KTRFJVRKVQROKS-UJCZHUFNSA-N

General description

Biliverdin (BV) is considered as a near-infrared (NIR)-absorbing pigment, that is endogenic. It is a linear tetrapyrrolic intermediate of heme degradation (heme catabolism) to bilirubin. The bile of amphibia and birds contains only biliverdine, no bilirubin.

Application

Biliverdin hydrochloride has been used:
  • as a treatment to J774A.1 cells to study its role in alternation of beta (β)-endorphins level
  • as a commercial standard for the detection of biliverdin in mouse unstable and stable plaques tissues using liquid chromatography-tandem mass spectrometry (LC-MS-MS)
  • to add to Fucci-containing cells to improve the mIFP signal
  • to examine its impact on infrared fluorescent protein (iRFP) in some retinal preparations

Biochem/physiol Actions

Biliverdin (BV) is an excellent anti-inflammatory agent that protects against lipopolysaccharides (LPS)-induced lung injury in mouse endothelial cells. It aids protection in polymicrobial sepsis. Biliverdin has cytoprotective effects on rat livers and protects from prolonged ischemia and reperfusion injury. It also shows antioxidant activities.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biological implications of heme metabolism.
Sassa, S.
Journal of Clinical Biochemistry and Nutrition, 38, 138-155 (2006)
Nader G Abraham et al.
Pharmacological reviews, 60(1), 79-127 (2008-03-08)
This review is intended to stimulate interest in the effect of increased expression of heme oxygenase-1 (HO-1) protein and increased levels of HO activity on normal and pathological states. The HO system includes the heme catabolic pathway, comprising HO and
Constantino Fondevila et al.
Hepatology (Baltimore, Md.), 40(6), 1333-1341 (2004-11-27)
Heme oxygenase (HO-1) provides a cellular defense mechanism during oxidative stress and catalyzes the rate-limiting step in heme metabolism that produces biliverdin (BV). The role of BV and its potential use in preventing ischemia/reperfusion injury (IRI) had never been studied.
Tracking multiple genomic elements using correlative CRISPR imaging and sequential DNA FISH
Guan J, et al.
Biophysical Journal, 112(6), 1077-1084 (2017)
Marking cells with infrared fluorescent proteins to preserve photoresponsiveness in the retina
Fyk-Kolodziej B, et al.
Biotechniques, 57(5), 245-253 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service