Skip to Content
Merck
All Photos(1)

Key Documents

278A-1

Sigma-Aldrich

Lysozyme Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (278A-14)
vial of 0.5 mL concentrate (278A-15)
bottle of 1.0 mL predilute (278A-17)
vial of 1.0 mL concentrate (278A-16)
bottle of 7.0 mL predilute (278A-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Anti-lysozyme stains myeloid cells, histiocytes, granulocytes, macrophages, and monocytes. It is an important marker that may demonstrate the myeloid or monocytic nature of acute leukemia. The restrictive nature of anti-lysozyme staining suggests that lysozyme may be synthesized predominantly in reactive histiocytes rather than in resting, unstimulated phagocytes. Anti-lysozyme may aid in the identification of histiocytic neoplasias, large lymphocytes, and classifying lymphoproliferative disorders.

Quality


IVD

IVD

IVD

RUO

Linkage

Lysozyme (polyclonal) Positive Control Slides, Product No. 278S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S T Yuen et al.
Histopathology, 32(2), 126-132 (1998-04-17)
The presence of lysozyme protein in some gastric adenomas and adenocarcinomas has been well documented. There have been relatively few studies investigating the presence of lysozyme in tumours of the large intestine and they show contrasting results. We aim to
H Kumamoto et al.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology, 30(4), 245-250 (2001-04-17)
Apoptotic cell death in granular cell ameloblastomas was examined by immunohistochemistry using anti-single-stranded DNA (ssDNA) antibody and transmission electron microscopy. Routinely prepared sections of granular cell ameloblastomas showed various quantities of granular cells with some apoptotic nuclear fragments. Immunoreactivity for
P Mörsky
Clinica chimica acta; international journal of clinical chemistry, 178(3), 327-336 (1988-12-30)
An immunoblotting technique was developed to detect human lysozyme and lysozyme complexes in body fluids. The unoccupied binding capacity of proteins was demonstrated by addition of surplus lysozyme. The sensitivity of immunoblotting to the free enzyme in human albumin solution
E Delaflor-Weiss et al.
Acta cytologica, 43(6), 1124-1130 (1999-12-01)
Granulocytic sarcoma of the uterine cervix is an unusual manifestation of acute myeloid leukemia, representing soft tissue masses of leukemic myeloblasts. An often misdiagnosed entity, it is often confused with other inflammatory or neoplastic conditions, including large cell lymphoma. A
L Krugliak et al.
American journal of hematology, 21(1), 99-109 (1986-01-01)
Blood and bone marrow samples from 20 individuals with reactive conditions and 26 cases of acute and chronic myeloid leukemias were tested for the presence of lysozyme, alpha-1-antitrypsin (alpha-1-AT), and alpha-1-antichymotrypsin (alpha-1-ACT). We compared the reactivity of samples in smears

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service