Skip to Content
Merck
All Photos(3)

Key Documents

H6254

Sigma-Aldrich

Hyaluronidase from sheep testes

Type V, lyophilized powder, ≥1,500 units/mg solid

Synonym(s):

Hyaluronate 4-glycanohydrolase, Hyaluronoglucosaminidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

sheep testis

Quality Level

type

Type V

form

lyophilized powder

specific activity

≥1,500 units/mg solid

mol wt

55 kDa

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Hyaluronidase is present in high amount in testis as it aids fertilization of the mammalian egg.[1] Mammalian hyaluronidase has a catalytic epidermal growth factor (EGF)-like domain and a C-terminal cysteine rich region.[2]

Application

Hyaluronidase has been used:
  • as a component of enzyme mix for the isolation of sertoli and germ cells from seminiferous tubules[3]
  • in the pre-treatment of deparaffinized adenomas sections for immunohistochemistry[4]
  • in the digestion of umbilical cord blood vessel for the generation of umbilical cord stromal stem cell lines[5]

Biochem/physiol Actions

Hyaluronidase from sheep testis is commercially used to depolymerise hyaluronic acid and chondroitin-sulphate in connective tissue and cartilage respectively.[1]
These enzymes randomly cleave β-N-acetylhexosamine-(1−4)-glycosidic bonds in hyaluronic acid, chondroitin, and chondroitin sulfates.

Unit Definition

One unit will degrade 0.75 μg of the polysaccharide hyaluronic acid per minute at pH 5.35 at 37 ºC (as measured by turbidimetric absorbance (λ: 600nm) when complexed with BSA after 45 minutes).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of human hyaluronidase-1, a hyaluronan hydrolyzing enzyme involved in tumor growth and angiogenesis
Chao KL, et al.
Biochemistry, 46(23), 6911-6920 (2007)
Human umbilical cord stromal stem cell express CD10 and exert contractile properties
Farias VA, et al.
Placenta, 32(1), 86-95 (2011)
Isolation of Sertoli cells from adult rat testes: an approach to ex vivo studies of Sertoli cell function
Anway MD, et al.
Biology of Reproduction, 68(3), 996-1002 (2003)
Yan Liang et al.
Scientific reports, 7(1), 12148-12148 (2017-09-25)
Meniscus fibrochondrocytes (MFCs) may be the optimal cell source to repair non-healing meniscus injuries using tissue engineering strategies. In this study, we investigated the effects of mitotic divisions and oxygen tension on the plasticity of adult human MFCs. Our assessment
Samia Rahman et al.
Frontiers in cell and developmental biology, 9, 703038-703038 (2021-08-13)
Articular cartilage of the knee joint is avascular, exists under a low oxygen tension microenvironment, and does not self-heal when injured. Human infrapatellar fat pad-sourced mesenchymal stem cells (IFP-MSC) are an arthroscopically accessible source of mesenchymal stem cells (MSC) for

Questions

  1. Mit wie viel Aqua dest muss das Hyaluronidase Typ V 500mg rekonstruiert werden?

    1 answer
    1. Hyaluronidase is soluble in water, however most aqueous buffers will be suitable. For testing purposes the bovine enzyme is reconstituted in 20 mM sodium phosphate buffer, with 77 mM sodium chloride, 0.01% BSA, pH 7.0. Solutions are stable up to 1 month refrigerated at 4 deg. C. For long term storage, maintain a concentration of >1 mg/mL, aliquot and freese at - 20 deg. C. Avoid repeated freeze/thaw cycles.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service