Skip to Content
Merck
All Photos(1)

Documents

171M-9

Sigma-Aldrich

CD71 (MRQ-48) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-48, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (171M-94)
vial of 0.5 mL concentrate (171M-95)
bottle of 1.0 mL predilute (171M-97)
vial of 1.0 mL concentrate (171M-96)
bottle of 7.0 mL predilute (171M-98)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

bone marrow

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

General description

CD71, also known as the transferrin receptor, is a membrane glycoprotein that mediates the uptake of iron from transferrin for hemoglobin synthesis in erythroid cells. Early erythroid precursors and erythroblasts contain the highest mass of transferrin receptors, and expression is lost as these cells cease hemoglobin synthesis and mature into erythrocytes. Therefore, anti-CD71 is a useful marker for highlighting erythroid precursors in normal and neoplastic specimens.

Quality


IVD

IVD

IVD

RUO

Linkage

CD71 Positive Control Slides, Product No. 171S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Ponka et al.
The international journal of biochemistry & cell biology, 31(10), 1111-1137 (1999-12-03)
The transferrin receptor is a membrane glycoprotein whose only clearly defined function is to mediate cellular uptake of iron from a plasma glycoprotein, transferrin. Iron uptake from transferrin involves the binding of transferrin to the transferrin receptor, internalization of transferrin
J Lesley et al.
Cellular immunology, 83(1), 14-25 (1984-01-01)
We have used a monoclonal antibody against the murine transferrin receptor to study the expression of the transferrin receptor on the hematopoietic progenitor cells (BFU-E, CFU-E, and CFU-C) present in mouse bone marrow. Elutriation and cell-sorting data are consistent with
C Sieff et al.
Blood, 60(3), 703-713 (1982-09-01)
Human bone marrow cells were separated on a fluorescence activated cell sorter (FACS) according to their binding of a series of monoclonal antibodies; the positive and negative fractions were cloned for erythroid burst and colony-forming units (BFU-E and CFU-E) and
T Nakahata et al.
Leukemia & lymphoma, 13(5-6), 401-409 (1994-05-01)
This review summarizes the changes in cell surface antigen expression during proliferation and differentiation of human erythroid progenitors. The content is based on our experimental data obtained from complement-mediated cytotoxicity assays against hematopoietic progenitors and a combined technique of sequential
Derek K Marsee et al.
American journal of clinical pathology, 134(3), 429-435 (2010-08-19)
Accurate analysis of the erythroid lineage is essential in evaluating bone marrow biopsy specimens and can be particularly challenging in the setting of dyserythropoiesis. Transferrin receptor (CD71) mediates the uptake of transferrin-iron complexes and is highly expressed on the surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service