Skip to Content
Merck
All Photos(1)

Key Documents

13264

Sigma-Aldrich

Solanidine

≥97.0% (HPLC)

Synonym(s):

(3β)-Solanid-5-en-3-ol, Solatubine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H43NO
CAS Number:
Molecular Weight:
397.64
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥97.0% (HPLC)

form

powder or crystals

technique(s)

HPLC: suitable

storage temp.

room temp

SMILES string

C[C@H]1CC[C@@H]2[C@@H](C)[C@H]3[C@H](C[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5CC[C@]34C)N2C1

InChI

1S/C27H43NO/c1-16-5-8-23-17(2)25-24(28(23)15-16)14-22-20-7-6-18-13-19(29)9-11-26(18,3)21(20)10-12-27(22,25)4/h6,16-17,19-25,29H,5,7-15H2,1-4H3/t16-,17+,19-,20+,21-,22-,23+,24-,25-,26-,27-/m0/s1

InChI key

JVKYZPBMZPJNAJ-OQFNDJACSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A poisonous steroid alkaloid derived from cholesterol and L-Arginine that is present in plants of the Solanaceae family. Solanidine is an inhibitor of serum cholinesterases in humans.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuyuki Kozukue et al.
Journal of agricultural and food chemistry, 56(24), 11920-11928 (2008-12-05)
Steroidal glycoalkaloids are naturally occurring, secondary plant metabolites that are found in foods, including potatoes and tomatoes. Their content in plants is controlled by both genetic and environmental factors. Glycoalkaloid profiles can be passed to progenies during breeding and hybridization
M Friedman et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 30(8), 689-694 (1992-08-01)
To assess whether reported toxicities of potato-derived glycoalkaloids could be the result of interactions with cellular DNA, the genotoxic effects of alpha-solanine, alpha-chaconine and solanidine were studied, using the Ames test (Salmonella strains TA98 and TA100), the mouse peripheral blood
J Laurila et al.
Journal of agricultural and food chemistry, 47(7), 2738-2742 (1999-12-20)
A combined derivatization method for gas chromatographic/mass spectrometric (GC/MS) analysis of steroidal glycoalkaloid (SGA) aglycons was developed using both trimethylsilylation and pentafluoropropionylation. In comparison with underivatized or only silylated aglycons, the new technique produces more specific and abundant fragmentation for
L F Neville et al.
Journal of neuroscience research, 27(4), 452-460 (1990-12-01)
The "atypical" allelic variant of human butyrylcholinesterase (BuChE) can be characterized by its failure to bind the local anesthetic dibucaine, the muscle relaxant succinylcholine, and the naturally occurring steroidal alkaloid solanidine, all assumed to bind to the charged anionic site
D R Driedger et al.
Journal of agricultural and food chemistry, 48(4), 1135-1139 (2000-04-25)
Solution-phase immunoassays are typically faster and more precise than ELISAs. This research developed a solution-phase for the immunoassay of potato glycoalkaloids (GAs) based on quantification by capillary electrophoresis (CE) with laser-induced fluorescence (LIF) detection. Solanidine coupled to 4'-(aminomethyl)fluorescein and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service