Skip to Content
Merck
All Photos(1)

Key Documents

12353

Supelco

Benzoic acid

reference material for titrimetry, certified by BAM, >99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
Beilstein:
636131
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39023903
PubChem Substance ID:
NACRES:
NA.24

grade

reference material

vapor density

4.21 (vs air)

vapor pressure

10 mmHg ( 132 °C)

Assay

>99.5%

autoignition temp.

1061 °F

quality

certified by BAM

technique(s)

titration: suitable

bp

249 °C (lit.)

mp

121-125 °C (lit.)

solubility

water: soluble (2.9 g/l at 25 °C)

density

1.32 g/cm3 at 20 °C

application(s)

environmental

format

neat

SMILES string

OC(=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoic acid is a volumetric standard produced under rigorous conditions and qualified with the highest possible precision. Its content determination and homogeneity assessment are performed by volumetric neutralization as the primary analysis method.

Application

It is used as a reference material for the standardization of volumetric solutions in alkalimetric titrations.

Features and Benefits

  • Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • Traceable to NIST SRM
  • High-quality offering accurate titer determinations
  • Accompanied by a certificate of analysis (CoA)

Analysis Note

Exact concentration, expiry date and detailed information on certification can be found on the certificate and certification report, included in each package. Content and expiry date on label.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allen J Duplantier et al.
Journal of medicinal chemistry, 52(11), 3576-3585 (2009-05-15)
3-Hydroxyquinolin-2(1H)-one (2) was discovered by high throughput screening in a functional assay to be a potent inhibitor of human DAAO, and its binding affinity was confirmed in a Biacore assay. Cocrystallization of 2 with the human DAAO enzyme defined the
S Nacht et al.
Journal of the American Academy of Dermatology, 4(1), 31-37 (1981-01-01)
The transepidermal penetration and metabolic disposition of 14C-benzoyl peroxide were assessed in vitro (excised human skin) and in vivo (rhesus monkey). In vitro, the benzoyl peroxide penetrated into the skin, through the stratum corneum or the follicular openings, or both
Copper-mediated C-H/C-H biaryl coupling of benzoic acid derivatives and 1,3-azoles.
Mayuko Nishino et al.
Angewandte Chemie (International ed. in English), 52(16), 4457-4461 (2013-03-21)
Wanchun Xiang et al.
ChemSusChem, 6(2), 256-260 (2013-01-25)
Sensing the sun: Incorporation of a cyanomethyl benzoic acid electron acceptor into donor-π-acceptor sensitizers for dye-sensitized-solar cell is shown to lead to devices with improved conversion efficiency when compared with more widely used cyanoacetic acid acceptor.
Ly Dieu Tran et al.
Journal of the American Chemical Society, 134(44), 18237-18240 (2012-10-30)
An auxiliary-assisted, copper catalyzed or promoted sulfenylation of benzoic acid derivative β-C-H bonds and benzylamine derivative γ-C-H bonds has been developed. The method employs disulfide reagents, copper(II) acetate, and DMSO solvent at 90-130 °C. Application of this methodology to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service