Skip to Content
Merck
All Photos(1)

Key Documents

10831

Sigma-Aldrich

Histopaque®-1083

sterile-filtered, density: 1.083

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

sterility

sterile-filtered

form

liquid

shelf life

Expiry date on the label.

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

density

1.083

application(s)

hematology
histology

storage temp.

2-8°C

General description

Histopaque®-1083 is a solution containing polysucrose and sodium diatrizoate, adjusted to a relative density of 1.083 g/mL. It facilitates the recovery of viable mononuclear cells from small volumes of blood of rats, mice, and other small mammals.

Application

Histopaque®-1083 has been used for the isolation of mononuclear cells and leukocytes from rat peripheral blood.

Features and Benefits

Histopaque®-1083 offers numerous advantages, such as:

  • Optimal recovery of viable cells
  • Permits selective separation of blood cell types
  • Prevents cell distortion
  • Preserves cell viability
  • Minimal extraneous cell interference
  • Reproducible performance from batch to batch
  • Sterile-filtered and endotoxin-tested
  • Stable at 2-8°C for at least 2 years when protected from light

Principle

Anticoagulated blood is layered onto HISTOPAQUE1083. During centrifugation, erythrocytes are aggregated by polysucrose and rapidly sediment; whereas, lymphocytes and other mononuclear cells remain at the plasma-HISTOPAQUE 1083 interface. Erythrocytes pellet to the bottom of the centrifuge tube.

Legal Information

Histopaque is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer J Chung et al.
The Journal of thoracic and cardiovascular surgery, 159(5), 1825-1835 (2019-07-30)
Extracellular vesicles (EVs) are promising therapeutics for cardiovascular disease, but poorly-timed delivery might hinder efficacy. We characterized the time-dependent response to endothelial progenitor cell (EPC)-EVs within an injectable shear-thinning hydrogel (STG+EV) post-myocardial infarction (MI) to identify when an optimal response
Gangduo Wang et al.
Toxicology and applied pharmacology, 237(2), 188-195 (2009-04-01)
Even though reactive oxygen and nitrogen species (RONS) are implicated as mediators of autoimmune diseases (ADs), little is known about contribution of protein oxidation (carbonylation and nitration) in the pathogenesis of such diseases. The focus of this study was, therefore
Bella S Guerrouahen et al.
Pediatric blood & cancer, 53(6), 1132-1135 (2009-06-02)
Tyrosine kinase inhibitors, such as imatinib, have dramatically improved the outcomes for patients with selected cancers. For imatinib, western blotting of phospho-CrkL was an insensitive, indirect, and descriptive method to determine drug efficacy. Greater use of targeted therapies should involve
Qiyue Hu et al.
Scientific reports, 8(1), 7675-7675 (2018-05-18)
Interleukin-15 (IL-15) can promote both innate and adaptive immune reactions by stimulating CD8+/CD4+ T cells and natural killer cells (NK) while showing no effect in activating T-regulatory (Treg) cells or inducing activation-associated death among effector T cells and NK cells.
Ahmad J H Albaghdadi et al.
Scientific reports, 9(1), 6528-6528 (2019-04-27)
Polycystic ovary syndrome (PCOS) is characterized by failure of ovulation and is associated with obesity and chronic inflammation. Recent evidence suggests that anomalous activation of ovarian macrophages and numerical and functional deficits in the Th17 (CD4+IL17A+) and the CD4+CD25+CD127low Tregs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service