100-500
Microvascular Endothelial Cell Basal Medium (500ml)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.71
Recommended Products
form
liquid
Quality Level
shelf life
1 yr
manufacturer/tradename
Cell Applications, Inc
shipped in
wet ice
storage temp.
2-8°C
General description
Add Growth Supplement before use
Quality
Each lot was tested for the ability to support plating, spreading and proliferation
Preparation Note
contained in protocol
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Matthew R Preimesberger et al.
Journal of inorganic biochemistry, 177, 171-182 (2017-10-03)
The cyanobacterium Synechococcus sp. PCC 7002 produces a monomeric hemoglobin (GlbN) implicated in the detoxification of reactive nitrogen and oxygen species. GlbN contains a b heme, which can be modified under certain reducing conditions. The modified protein (GlbN-A) has one
Jayanta Kumar Patra et al.
Molecules (Basel, Switzerland), 20(7), 12093-12113 (2015-07-07)
Laminaria japonica L. is among the most commonly consumed seaweeds in northeast Asia. In the present study, L. japonica essential oil (LJEO) was extracted by microwave-hydrodistillation and analyzed by gas chromatography and mass spectroscopy. LJEO contained 21 volatile compounds, comprising
Zhan Ye et al.
Food research international (Ottawa, Ont.), 111, 281-290 (2018-07-17)
The digestion fates of different edible oils are different. The objective of this study was to understand the influences of lipid composition on their digestion fates, and investigate the roles of bile salts (BS) played in emulsified lipid system (whey
Zhan Ye et al.
Food & function, 10(3), 1490-1503 (2019-02-21)
The objective of the present study was to investigate the connections between lipid compositions and the digestion and absorption differences of different lipids. Five typical edible oils (palm oil, PO; leaf lard oil, LO; rapeseed oil, RO; sunflower oil, SO;
Jie Gong et al.
Shock (Augusta, Ga.), 44(4), 371-380 (2015-07-22)
Acute lung injury (ALI) is characterized by lung inflammation and diffuse infiltration of neutrophils. Neutrophil apoptosis is recognized as an important control point in the resolution of inflammation. Maresin 1 (MaR1) is a new docosahexaenoic acid-derived proresolving agent that promotes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service