09962
Diastase from Aspergillus oryzae
powder, white, amylase ≥3500 U/g
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder
amylase activity
≥3500 U/g
color
white
storage temp.
2-8°C
Related Categories
Application
Diastase from Aspergillus oryzae has been used:
- to distinguish sulphomucins and sialomucins
- to discriminate acidic mucins from neutral mucins in the lingual salivary glands
- as a component of the BioVision amylase activity assay buffer solution for in vitro and ex vivo degradation assays
Biochem/physiol Actions
Diastase catalyzes the digestion of glycogen. It is useful as a reducing and stabilizing agent in the aqueous synthesis of gold nanoparticles (AuNPs).
Unit Definition
1 U corresponds to the amount of enzyme which liberates 1 μmol maltose per minute at pH 6.0 and 25 °C (starch acc. to Zulkowsky, Cat. No. 85642, as substrate)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ultrastructure of the tongue and histochemical features of the lingual salivary glands in buzzards
Bozkurt EU, et al.
Turkish Journal of Veterinary and Animal Sciences, 42, 161-167 null
Inheritable Forms of Liver Disease
Bacon BR
Liver, 243-243 (2014)
Kesarla Mohan Kumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 116, 539-545 (2013-08-27)
A facile rapid green eco-friendly method to synthesize gold nanoparticles (Au NPs) of tunable size using aqueous Terminalia arjuna fruit extracts has been demonstrated herein. Formation of Au NPs was confirmed by Surface Plasmon Resonance (SPR) study at 528 nm
Grant L Austin et al.
Molecular pharmaceutics, 16(9), 3791-3801 (2019-07-23)
Lafora disease (LD) is a fatal juvenile epilepsy characterized by the accumulation of aberrant glucan aggregates called Lafora bodies (LBs). Delivery of protein-based therapeutics to the central nervous system (CNS) for the clearance of LBs remains a unique challenge in
F Başak et al.
Folia morphologica, 76(3), 348-354 (2017-02-16)
In this study, morphological characteristics of the canary tongue were examined macroscopically and histologically besides using scanning electron microscopy. Furthermore, histochemical features of the lingual salivary glands of the canary were also examined. The results suggest that the tongue of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service