Skip to Content
Merck
All Photos(1)

Key Documents

A4582

Supelco

α-Amylase from Bacillus licheniformis

suitable for determination of starch (Kit STA-20)

Synonym(s):

1,4-α-D-Glucan-glucanohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12164500
eCl@ss:
32160410
NACRES:
NA.84

form

liquid

Quality Level

analyte chemical class(es)

sugars

technique(s)

photometry: suitable

suitability

suitable for determination of starch (Kit STA-20)

application(s)

food and beverages
general analytical

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

α-Amylase belongs to glycosyl hydrolase family 13.

Application

α-Amylase from Bacillus licheniformis has been used in two-step hydrolysis for enzymatic saccharification of dried biomass powder. It has also been used to analyse the starch concentration by an enzymatic method.

Biochem/physiol Actions

α-Amylase, the digestive enzyme in saliva has several significance in industries like, food, pharmaceutical, textile and paper. It hydrolyses the linear chain of amylose to form glucose molecules.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SALIVARY GLANDS AND OESOPHAGUS
Digestive Disease Pathology (2010)
The response of rice grain quality to ozone exposure during growth depends on ozone level and genotype
Frei M, et al.
Environmental Pollution (Barking, Essex : 1987), 163, 199- 206 (2012)
Growing Lemna minor in agricultural wastewater and converting the duckweed biomass to ethanol
Ge X, et al.
Bioresource Technology, 124, 485- 488 (2012)
An overview of the microbial alpha-amylase family
Reddy NS, et al.
African Journal of Biotechnology, 2(12), 645- 648 (2003)
Changjiang Yu et al.
Biotechnology for biofuels, 10, 167-167 (2017-07-04)
Duckweed is considered a promising source of energy due to its high starch content and rapid growth rate. Starch accumulation in duckweed involves complex processes that depend on the balanced expression of genes controlled by various environmental and endogenous factors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service