Skip to Content
Merck

Skip To

08014

Micro particles, magnetic, streptavidin coated

10 μm particle size, std dev <0.5 μm

Synonym(s):

Polystyrene beads, magnetic, streptavidin coated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous solution

Quality Level

composition

iron oxide, ≥20%

concentration

1% (solids)

particle size

10 μm std dev <0.5 μm

storage temp.

2-8°C

General description

Micro Particles or Latex Beads, commonly known as polystyrene microparticles, are negatively charged colloidal particles. The microparticles are produced through a polymerization reaction of styrene under controlled conditions. The conducive environment of the reaction induces spontaneous coalescent bead formation. The biotin-streptavidin system is the strongest noncovalent biological interaction known, having a dissociation constant, K(d), in the order of 4x10(-14) M.[1] The beads are magnetic in nature and coated with streptavidin for enhanced binding. Micro Particles or Latex beads are supplied in aqueous solutions composed of polymer particles with water. Polystyrene′s refractive index at 589 nm is 1.5905 and 1.602 at 486 nm.
Superparamagnetic polystyrene particles

Application

Easily conjugated with biotin, making them useful for several detection assays,.
Micro Particles or Latex Beads is used for a wide variety of applications like:
  • Electron Microscopy.
  • Cell Counter Calibration.
  • Immunodiagnostics, especially for latex agglutination tests, and
  • Phagocytosis Research.
Microparticles with streptavidin are easily conjugated with biotin for use in various detection assays

Analysis Note

Binding capacity for FITC-Biotin: >300 pmol/mg

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
396894953249664
concentration

1% (solids)

concentration

5% solids

concentration

1% (solids)

concentration

5% solids

particle size

10 μm std dev <0.5 μm

particle size

5 μm std dev <0.5 μm

particle size

1 μm std dev <0.1 μm

particle size

10 μm std dev <0.5 μm

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

form

aqueous solution

form

aqueous solution

form

aqueous solution

form

aqueous solution

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

composition

iron oxide, ≥20%

composition

iron oxide, ≥20%

composition

iron oxide, ≥20%

composition

iron oxide, ≥20%


Still not finding the right product?


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Anders Holmberg et al.
Electrophoresis, 26(3), 501-510 (2005-02-04)
The biotin-streptavidin system is the strongest noncovalent biological interaction known, having a dissociation constant, K(d), in the order of 4x10(-14) M. The strength and specificity of the interaction has led it to be one of the most widely used affinity
Rongrong Huang et al.
Autophagy, 13(10), 1722-1741 (2017-08-09)
Circular RNAs are a subclass of noncoding RNAs in mammalian cells; however, whether these RNAs are involved in the regulation of astrocyte activation is largely unknown. Here, we have shown that the circular RNA HIPK2 (circHIPK2) functions as an endogenous



Global Trade Item Number

SKUGTIN
08014-5ML04061838210463
08014-10ML04061838210456

Questions

1–2 of 2 Questions  
  1. What is the concentration of the beads (number/ml)?

    1 answer
    1. The estimated particle concentration is 1.3 x 10^7 per milliliter. This is a calculated value based on the following specifications:
      Polymer density: 1.47 g/cc
      Mean particle diameter: 10 um
      Particle concentration in percentage of aq. suspension: 10 mg/ml (1%).

      Helpful?

  2. Could you reveal the buffer composition for the buffer that the beads are supplied in?

    1 answer
    1. This product is prepared in water rather than a buffer solution. No surfactant has been added.

      Helpful?

Reviews

No rating value

Active Filters