Skip to Content
Merck

Skip To

06563

Methotrexate hydrate

≥99.0% (sum of enantiomers, HPLC)

Synonym(s):

4-Amino-10-methylfolic acid hydrate, L-4-Amino-N10-methylpteroylglutamic acid hydrate, L-Amethopterin hydrate, Antifolan hydrate, MTX hydrate, Methylaminopterin hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H22N8O5 · xH2O
CAS Number:
Molecular Weight:
454.44 (anhydrous basis)
PubChem Substance ID:
UNSPSC Code:
12352202
NACRES:
NA.77
EC Number:
200-413-8
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Level

assay

≥99.0% (sum of enantiomers, HPLC)

form

powder or crystals

optical activity

[α]/D +21.0±2.0°

impurities

≤0.1% sulfated ash

mp

185-204 °C

solubility

water: insoluble

storage temp.

−20°C

SMILES string

[H]O[H].CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C20H22N8O5.H2O/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30;/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27);1H2/t13-;/m0./s1

InChI key

FPJYMUQSRFJSEW-ZOWNYOTGSA-N

Gene Information

human ... DHFR(1719)

General description

Methotrexateis an analog of folate.[1] It shows anti-inflammatory effects throughseveral cellular mechanisms. The inhibition of dihydrofolate reductase reducesthe de novo synthesis of pyrimidines and purines, transmetylation ofphospholipids and proteins, and formation of polyamines.[2]

Application

Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems.
Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems. Also effective in treatment of pyrimethamine-resistant Plasmodium vivax malaria parasites.
Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems. Also shows immunosuppressive effects in, e.g., rheumatoid arthritis.
Methotrexatehydrate has been used as a disease modifyinganti-rheumatic drug (DMARD) to study its effects on Ross River virus disease (RRVD) in mice models.

Other Notes

Folic acid antagonist. Potent inhibitor of dihydrofolate reductase.[3]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A6770M8407PHR1396
biological source

synthetic

biological source

synthetic (organic)

biological source

-

biological source

-

assay

≥99.0% (sum of enantiomers, HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

-

Gene Information

human ... DHFR(1719)

Gene Information

human ... DHFR(1719)

Gene Information

human ... DHFR(1719)

Gene Information

human ... DHFR(1719)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

form

powder or crystals

form

powder

form

powder

form

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Methotrexate hydrate


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 1B - STOT RE 1

target_organs

Liver,Bone marrow

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Methotrexate Treatment Causes Early Onset of Disease in a Mouse Model of Ross River Virus-Induced Inflammatory Disease through Increased Monocyte Production
Adam Taylor, et al.
PLoS ONE (2013)
Edwin S L Chan et al.
Nature reviews. Rheumatology, 6(3), 175-178 (2010-03-04)
Methotrexate remains a cornerstone in the treatment of rheumatoid arthritis and other rheumatic diseases. Folate antagonism is known to contribute to the antiproliferative effects that are important in the action of methotrexate against malignant diseases, but concomitant administration of folic
B N Cronstein
Rheumatic diseases clinics of North America, 23(4), 739-755 (1997-11-15)
Because of methotrexate's well-documented efficacy in the treatment of rheumatoid arthritis, it is important that we understand the mechanism of action of this drug. There are two biochemical mechanisms by which methotrexate may modulate inflammation: (1) promotion of adenosine release



Global Trade Item Number

SKUGTIN
821189100004022536475653
821189010004022536475646
T81108-500G04061837378904
T81108-2KG04061837378898
T81108-25G04061832886442
821189902504027269017323
06563-5MG04061838646392
06563-25MG04061838646385
06563-100MG04061838646378

Questions

Reviews

No rating value

Active Filters