Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.32
UNSPSC Code:
12352108
Pricing and availability is not currently available.
product line
BioReagent
assay
≥90% (coupling to amines)
mol wt
Mw 820 g/mol
manufacturer/tradename
ATTO-TEC GmbH
fluorescence
λex 663 nm; λem 684 nm in 0.1 M phosphate pH 7.0
suitability
suitable for fluorescence
storage temp.
−20°C
General description
Atto 665 is a new fluorescent label closely related to Atto 647N. It shows extraordinary high fluorescence quantum yield, excellent thermal and photo-stability, outstanding ozone resistance, and very little triplet formation. Atto 665 is a cationic (charge +1). The dye is moderately hydrophilic. The NHS-ester of the dye reacts easily with amino-groups of proteins and other bio-molecules. Since the amino group must be non protonated to be reactive, the pH of the reaction is recommended to be about 8.3. Prepare labeling solution of NHS-ester immediately before use by dissolving the vial content in anhydrous and amine-free DMF or DMSO.
find more information here
find more information here
Legal Information
This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.
1 of 1
This Item | |||
|---|---|---|---|
| suitability suitable for fluorescence | suitability suitable for fluorescence | suitability suitable for fluorescence | suitability suitable for fluorescence |
| fluorescence λex 663 nm; λem 684 nm in 0.1 M phosphate pH 7.0 | fluorescence - | fluorescence - | fluorescence - |
| assay ≥90% (coupling to amines) | assay ≥90% (HPLC), ≥90% (degree of coupling) | assay ≥90% (HPLC), ≥90% (degree of coupling) | assay ≥90% (HPLC) |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| product line BioReagent | product line BioReagent | product line BioReagent | product line - |
| mol wt Mw 820 g/mol | mol wt - | mol wt Mw 793 g/mol | mol wt - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 122432-1G | 04061825770581 |
| 04022-1MG | 04061823363693 |
