Skip to Content
Merck
All Photos(3)

Key Documents

S7757

Sigma-Aldrich

Sodium thiocyanate

reagent grade, 98-102% (titration)

Synonym(s):

Sodium isothiocyanate, Sodium rhodanate, Sodium rhodanide, Sodium sulfocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaSCN
CAS Number:
Molecular Weight:
81.07
Beilstein:
3594965
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Assay

98-102% (titration)

form

powder

pH

4.8 (20.1 °C, 1070 g/L)

mp

287 °C (dec.) (lit.)

SMILES string

[Na]SC#N

InChI

1S/CHNS.Na/c2-1-3;/h3H;/q;+1/p-1

InChI key

VGTPCRGMBIAPIM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Effect of Sodium Thiocyanate and Sodium Perchlorate on Poly(N-isopropylacrylamide) Collapse: This research investigates the effects of sodium thiocyanate on the collapse of poly(N-isopropylacrylamide). The study provides insights into the molecular interactions and thermodynamic behavior of this polymer in the presence of thiocyanate ions (Pica & Graziano, 2020).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sandra Romero-Steiner et al.
Clinical and diagnostic laboratory immunology, 12(9), 1029-1035 (2005-09-09)
Determination of antibody avidity measurements can be difficult in human serum depending on the population evaluated. We evaluated three approaches for the determination of antibody avidity for immunoglobulin G (IgG). These approaches were (i) elution of bound antibody with increasing
Vincent Ball et al.
Langmuir : the ACS journal of surfaces and colloids, 21(9), 4129-4137 (2005-04-20)
In this study, we investigate the buildup of PEI-(PSS-PAH)(n) polyelectrolyte multilayers at pH 7.4 in the presence of either NaCl or NaSCN as a supporting electrolyte. It appears that in the presence of increasing thiocyanate concentrations (from 0.1 to 0.5
S-F Chin et al.
Molecular pathology : MP, 56(5), 275-279 (2003-09-30)
To describe a robust pretreatment protocol for preparing paraffin wax embedded tissues on tissue microarrays for fluorescence in situ hybridisation (FISH). The newly developed pretreatment protocol described here was compared with the commonly used sodium thiocyanate based protocol and two
Edyta Glogowska et al.
Blood cells, molecules & diseases, 44(4), 243-251 (2010-03-17)
Historically, the anion transport through the human red cell membrane has been perceived to be mediated by Band 3, in the two-component concept with the large electroneutral anion exchange accompanied by the conductance proper, which dominated the total membrane conductance.
Z G Peng et al.
Colloids and surfaces. B, Biointerfaces, 35(3-4), 169-174 (2004-07-21)
Adsorption and desorption of lysozyme on nano-sized magnetic particles and its conformational change were studied in this work. Adsorption of lysozyme on nano-sized magnetic particles (Fe(3)O(4)) was carried out at different pH. Maximum adsorption of lysozyme (4.65 mg/m2) occurred at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service