Skip to Content
Merck
All Photos(2)

Key Documents

M79603

Sigma-Aldrich

NMP

ReagentPlus®, 99%

Synonym(s):

1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein:
106420
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
bp:
202 °C (lit.)
81-82 °C/10 mmHg (lit.)
vapor pressure:
0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

Product Name

1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%

vapor density

3.4 (vs air)

Quality Level

vapor pressure

0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

518 °F

expl. lim.

9.5 %

dilution

(for general lab use)

impurities

≤0.1% water (Karl Fischer)

color

APHA: ≤50

refractive index

n20/D 1.47 (lit.)

pH

7.7-10.0 (20 °C, 100 g/L)

bp

202 °C (lit.)
81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

SMILES string

CN1CCCC1=O

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Undergoes condensation reactions with arylthioacetamides and methyl salicylates. Versatile solvent for water miscible applications.

Other Notes

A Greener alternative is available for many NMP applications, Cyrene (807796)

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Chemical Communications, 1437-1437 (1994)
J. Mol. Catal., 85, L109-L109 (1993)
The Journal of Organic Chemistry, 58, 7277-7277 (1993)
U Isele et al.
Journal of pharmaceutical sciences, 83(11), 1608-1616 (1994-11-01)
This work describes the development of an organic solvent dilution method suitable for the large scale manufacturing of small unilamellar liposomes containing the water-insoluble photosensitizer zinc phthalocyanine in the monomeric state. N-Methyl pyrrolidone (NMP)/tert-butyl alcohol was selected as water miscible
David S Hewings et al.
Journal of medicinal chemistry, 54(19), 6761-6770 (2011-08-20)
Histone-lysine acetylation is a vital chromatin post-translational modification involved in the epigenetic regulation of gene transcription. Bromodomains bind acetylated lysines, acting as readers of the histone-acetylation code. Competitive inhibitors of this interaction have antiproliferative and anti-inflammatory properties. With 57 distinct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service