Skip to Content
Merck
All Photos(4)

Documents

B2507

Sigma-Aldrich

Barium hydroxide octahydrate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(OH)2 · 8H2O
CAS Number:
Molecular Weight:
315.46
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

Assay

≥98%

form

powder

pH

12.5 (20 °C, 50 g/L)

mp

78 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O[Ba]O

InChI

1S/Ba.10H2O/h;10*1H2/q+2;;;;;;;;;;/p-2

InChI key

ZUDYPQRUOYEARG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyzes the β-elimination of phosphoserine residues.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Mahendran et al.
Journal of agricultural and food chemistry, 56(19), 9269-9276 (2008-09-12)
The structural characteristics of the gum exudate of Acacia senegal (gum arabic) have been investigated by monitoring the composition and physicochemical properties before and after treatment with proteolytic enzyme and various alkaline systems. Molecular mass ( M w) and radius
K C Li et al.
Magnetic resonance imaging, 9(2), 141-150 (1991-01-01)
In vitro proton spectroscopy with line-width measurements and MR imaging were performed on various concentrations of commercially available single contrast (SC), double contrast, oral and rectal barium sulfate suspensions, as well as potassium sulfate, barium chloride, barium hydroxide, and 97%
T Irie et al.
Carbohydrate research, 192, 167-172 (1989-10-23)
The alkylation of cyclomalto-oligosaccharides (cyclodextrins, CDs) with dialkyl sulfate-barium hydroxide has been claimed to yield 2,6-di-O-alkyl derivatives. Re-investigation by plasma desorption-m.s. of the products of laboratory methylation of alpha CD, beta CD, or gamma CD and ethylation of beta CD
H Förster et al.
Der Anaesthesist, 46(12), 1071-1075 (1998-02-06)
The various components of commercial soda lime (sodium hydroxide, potassium hydroxide, calcium hydroxide, barium hydroxide) were studied in terms of their reactivity with sevoflurane at its boiling point (59 degrees C). A simple closed system, a reflux cooler, served as
Marshall B Dunning et al.
Anesthesiology, 106(1), 144-148 (2007-01-02)
Fires, explosions, and extreme heat production may occur when sevoflurane reacts with desiccated barium hydroxide lime. The identity of the flammable gas has not previously been published, although carbon monoxide, methanol, formaldehyde, and methyl formate have been identified in low

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service