Skip to Content
Merck
All Photos(4)

Key Documents

A2939

Sigma-Aldrich

Ammonium sulfate

BioXtra, ≥99.0%

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38030408
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
form:
powder or crystals
solubility:
H2O: 1 M, clear, colorless
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥99.0%

form

powder or crystals

impurities

Insoluble matter, passes filter test

ign. residue

<0.01% (as SO4)

loss

≤1.0% loss on drying, 110°C

pH

5.0-6.0 (20 °C, 1 M in H2O)

mp

>280 °C (dec.) (lit.)

solubility

H2O: 1 M, clear, colorless

density

1.77 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.001%
phosphate (PO43-): ≤0.0005%

cation traces

Al: ≤0.0005%
As: ≤0.0001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0001%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0002%
Fe: ≤0.0002%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Na: ≤0.005%
Ni: ≤0.0005%
Pb: ≤0.0002%
Sr: ≤0.0005%
Zn: ≤0.0001%

absorption

≤0.030 at 280 in H2O at 1 M
≤0.037 at 260 in H2O at 1 M

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium sulfate has been used as an additive in salting-out procedure to prepare the g-globulin fraction.[1][2] It is also used as a component in synthetic minimal dextrose (SD) liquid medium.[3]

Biochem/physiol Actions

Ammonium sulfate ((NH4)2SO4) is mainly used as a soil fertilizer. It is also used as a wood preservative. This inorganic salt plays a role in flame retardant chemicals.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of turkey coronavirus in commercial turkey poults in Brazil
Teixeira MCB, et al.
Avian Pathology : Journal of the W.V.P.A, 36(1), 29-33 (2007)
Industrial Inorganic Chemistry
Environmental Nanotechnology, Monitoring & Management (2017)
Metabolic labeling of yeast sphingolipids with radioactive D-erythro-[4, 5-3H] dihydrosphingosine
Karashima T, et al.
Bio-protocol, 3, e862-e862 (2013)
Immunohistochemical detection of metalloproteinase-9 (MMP-9), anti-oxidant like 1 protein (AOP-1) and synaptosomal-associated protein (SNAP-25) in the cerebella of dogs naturally infected with spontaneous canine distemper
Bregano L, et al.
Folia Histochemica Et Cytobiologica, 49(1), 41-48 (2011)
Simon Gareth Broadley et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 11), 1450-1459 (2012-10-24)
MshB, a zinc-based deacetylase, catalyses a step in the mycothiol biosynthetic pathway that involves the deacetylation of 1-O-(2-acetamido-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcNAc-Ins), via cleavage of an amide bond, to 1-O-(2-amino-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcN-Ins) and acetate. In this study, MshB was expressed, purified and crystallized. A

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service