Skip to Content
Merck
All Photos(4)

Key Documents

A4514

Sigma-Aldrich

Ammonium chloride

ReagentPlus®, ≥99.5%

Synonym(s):

Salmiac

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
Beilstein:
4371014
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38050207
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
powder or crystals
solubility:
water: soluble 100 mg/mL

grade

reagent

Quality Level

Agency

suitable for SM 5210

vapor density

1.9 (vs air)

vapor pressure

1 mmHg ( 160.4 °C)

product line

ReagentPlus®

Assay

≥99.5%

form

powder or crystals

pH

4.5-5.5 (20 °C, 50 g/L)

mp

340 °C (subl.) (lit.)

solubility

water: soluble 100 mg/mL

SMILES string

N.Cl

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium chloride is commonly used as fertilizer. It can be synthesized by neutralizing ammonia with hydrochloric acid. It shows high vapor pressure at higher temperature. It is employed as an electrolyte during the production of drycell batteries. Its crystal structure has been analyzed by powder and single crystal neutron diffraction methods. Ammonium chloride shows change in some of the physical characteristics on its addition to a high carbonate containing unfired and fired illitic shale.

Application

Ammonium chloride may be used as a catalyst in the preparation of:
  • 3,4-dihydropyrimidin- 2-(1H)-ones
  • diindolylmethanes
  • 2-arylbenzothiazoles derivatives
  • 4(3H)-quinazolinones

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammonium Chloride-Catalyzed One-Pot Synthesis of 4(3H)-Quinazolinones Under Solvent-Free Conditions.
Huang G, et al.
Synthetic Communications, 44(12), 1786-1794 (2014)
Ammonium Compounds.
Weston CW, et al.
Kirk-Othmer Encyclopedia of Chemical Technology (2003)
Ammonium chloride; as a mild and efficient catalyst for the synthesis of some 2-arylbenzothiazoles and bisbenzothiazole derivatives.
Maleki B and Salehabadi H.
European Journal of Organic Chemistry, 1(4), 377-380 (2010)
Gowariker V, et al.
The Fertilizer Encyclopedia, 42-43 (2009)
Ammonium chloride catalyzed one-pot synthesis of diindolylmethanes under solvent-free conditions.
Azizian J, et al.
Catalysis Communications, 8(7), 1117-1121 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service