Skip to Content
Merck
All Photos(5)

Documents

213330

Sigma-Aldrich

Ammonium chloride

ACS reagent, ≥99.5%

Synonym(s):

Salmiac

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
Beilstein:
4371014
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38050207
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

suitable for SM 5210

vapor density

1.9 (vs air)

vapor pressure

1 mmHg ( 160.4 °C)

Assay

≥99.5%

form

powder, crystals or chunks

impurities

≤0.005% insolubles

ign. residue

≤0.01%

pH

4.5-5.5 (25 °C, 5%)

mp

340 °C (subl.) (lit.)

anion traces

phosphate (PO43-): ≤2 ppm
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.001%
Fe: ≤2 ppm
Mg: ≤5 ppm
heavy metals: ≤5 ppm (by ICP)

SMILES string

N.Cl

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crystals of ammonium chloride (NH4Cl) belong to the cubic crystal system. It can be synthesized by reacting HCl with aqueous ammonia solutions. NH4Cl is formed as a by-product during soda manufactured by Solvay process.

Application

Ammonium chloride (NH4Cl) has been used as one of the components of red blood cell (RBC) lysis buffer for use in various studies. It may be used as a catalyst in the preparation 3,4-dihydropyrimidin-2-(1H)-ones.
Inorganic ammonium ions (NH4+) effectively catalyze the aldol condensation of carbonyl compounds. It serves as a source of nitrogen during the preparation of trimethylamine hydrochloride (TMA·HCl).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

514502
CorpBase ID (for auto-filling citation data) null
Immune Cell Isolation from Mouse Femur Bone Marrow.
Liu X and Quan N.
Bio-protocol, 5(20) (2015)
Mark E Kusek et al.
Journal of visualized experiments : JoVE, (83)(83), e50823-e50823 (2014-01-17)
Mucosal surfaces serve as protective barriers against pathogenic organisms. Innate immune responses are activated upon sensing pathogen leading to the infiltration of tissues with migrating inflammatory cells, primarily neutrophils. This process has the potential to be destructive to tissues if
Selective Synthesis of Trimethylamine by Catalytic N-Methylation of Ammonia and Ammonium Chloride by utilizing Carbon Dioxide and Molecular Hydrogen.
Beydoun K, et al.
ChemCatChem, 8(1), 135-138 (2016)
Romina Frare et al.
FEMS microbiology letters, 367(23) (2020-11-27)
Nitrogen is a most important nutrient resource for Escherichia coli and other bacteria that harbor the glnKamtB operon, a high-affinity ammonium uptake system highly interconnected with cellular metabolism. Although this system confers an advantage to bacteria when growing under nitrogen-limiting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service