Skip to Content
Merck
All Photos(7)

Documents

791733

Sigma-Aldrich

Potassium acetate

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOK
CAS Number:
Molecular Weight:
98.14
Beilstein:
3595449
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
anhydrous

Quality Level

vapor pressure

<0.0000001 hPa ( 25 °C)

product line

Redi-Dri

Assay

≥99.0%

form

powder

quality

free-flowing

impurities

≤0.005% insolubles

pH

6.5-9.0 (25 °C, 5%)

density

1.57 g/cm3 at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.003%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.005%
Fe: ≤5 ppm
Mg: ≤0.002%
Na: ≤0.03%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[K+].CC([O-])=O

InChI

1S/C2H4O2.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

SCVFZCLFOSHCOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium acetate (KOAc) is the potassium salt of acetic acid. It is commonly used as a base in organic synthesis for various reactions, which include Lossen rearrangement of hydroxamic acids, E2 eliminations, deprotonations of alcohols, transition metal-catalyzed reactions, and fragmentation reactions. Additionally, KOAc is also used as a nucleophilic catalyst for addition and polymerization reactions.

Application

Potassium acetate can be used as an activating agent for the activation of carbon materials.

Features and Benefits

  • Increased efficiency – Eliminates time and effort of loosening hardened powders      
  • Safety – Eliminates tools of force used to break up clumps
  • Ecology – Reduces waste as it does not stick to the packaging
  • Economy – Faster preparation and solvation increase productivity and reduce costs     
  • Assured Quality – Excellent, expert-tested quality with no anti-caking agents      
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osvaldo P de Melo Neto et al.
RNA biology, 12(11), 1209-1221 (2015-09-05)
The eukaryotic initiation factor 4E (eIF4E) recognizes the mRNA cap structure and, together with eIF4G and eIF4A, form the eIF4F complex that regulates translation initiation in eukaryotes. In trypanosomatids, 2 eIF4E homologues (EIF4E3 and EIF4E4) have been shown to be
J Guitard et al.
Journal of clinical microbiology, 53(5), 1655-1661 (2015-03-13)
Candida inconspicua and Candida (Pichia) norvegensis are two emerging pathogenic species that exhibit reduced susceptibility to azole derivatives. Conventional (biochemical) approaches do not readily differentiate between the two species. The first aim of this work was to analyze the performance
Ya-Fang Huang et al.
Antioxidants & redox signaling, 23(12), 973-984 (2015-04-14)
To elucidate the role of reactive oxygen species (ROS) in arthritis and to identify targets of arthritis treatment in conditions with different levels of oxidant stress. Through establishing an arthritis model by injecting arthritogenic serum into wild-type and NADPH oxidase
Jun Wei Pek et al.
The Journal of cell biology, 211(2), 243-251 (2015-10-28)
Stable intronic sequence RNAs (sisRNAs) have been found in Xenopus tropicalis, human cell lines, and Epstein-Barr virus; however, the biological significance of sisRNAs remains poorly understood. We identify sisRNAs in Drosophila melanogaster by deep sequencing, reverse transcription polymerase chain reaction
Andreas Gietl et al.
Nucleic acids research, 42(10), 6219-6231 (2014-04-20)
During transcription initiation, the promoter DNA is recognized and bent by the basal transcription factor TATA-binding protein (TBP). Subsequent association of transcription factor B (TFB) with the TBP-DNA complex is followed by the recruitment of the ribonucleic acid polymerase resulting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service