Skip to Content
Merck
Potassium iodide BioXtra, ≥99.0%

Potassium iodide

Potassium iodide BioXtra, ≥99.0%
Synonym(s):
Potassium iodide
Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Special Grade
Technique
Showing 1-19 of 19
Compare
Product Number
Description
Pricing