Skip to Content
Merck
All Photos(1)

Documents

676764

Sigma-Aldrich

Tetrahydrofuran

≥99.0%, ACS reagent, contains 250 ppm BHT as inhibitor, suitable for HPLC

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
NACRES:
NA.07

product name

Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor

grade

ACS reagent

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

610 °F

contains

250 ppm BHT as inhibitor

expl. lim.

1.8-11.8 %

technique(s)

HPLC: suitable

impurities

≤0.015% peroxide (as H2O2)
≤0.05% water

evapn. residue

≤0.03%

color

APHA: ≤20

refractive index

n20/D 1.407 (lit.)

pH

~7

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofuran (THF) is a heterocyclic compound (cyclic ether). It is colorless, has low viscosity, and good solubility in a wide range of solvents. It is widely used as a solvent in organic synthesis, being very popular in reactions with organometallic compounds and Grignard reagents. Due to organic peroxides formation on long term storage, THF is usually stabilized by adding butylated hydroxytoluene (BHT). BHT removes the free radicals required for the peroxide formation.

Application

Tetrahydrofuran (THF) is used as a solvent in the following processes:
  • Organic synthesis
a) Grignard
b) Organometallic compounds
c) Reformatsky
d) Lithiation
e) Hydride reduction
f) Metal-catalyzed coupling (Heck, Stile, Suzuki)
g) Lewis acid mediated reactions
  • Crystallization
  • Polymerization. Ex. RAFT (Reversible Addition-Fragmentation Chain Transfer) polymerization of p-acetoxystyrene
  • Coatings
  • As an O-donor ligand to form coordination complexes
  • As mobile phase solvent in high-performance liquid chromatography

Features and Benefits

ACS solvents meet or exceed the high standards of the American Chemical Society (ACS) ,with test specifications that are specialized to every compound. According to the American Chemical Society, ACS reagent grade implies that it is a substance of sufficient purity to be used in most chemical analyses or reactions.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation of tetrahydrofuran to butyrolactone catalyzed by iron-containing clay.
Ausavasukhi A and Sooknoi T.
Green Chemistry, 17(1), 435-441 (2015)
Novel Diacetylinic Aryloxysilane Polymers: A New Thermally Cross-Linkable High Temperature Polymer System.
Drake K, et al.
Macromolecules, 46(11), 4370-4377 (2013)
An efficient and economical process for lignin depolymerization in biomass-derived solvent tetrahydrofuran.
Long J, et al.
Bioresource Technology, 154, 10-17 (2014)
1,3 Dioxolane versus tetrahydrofuran as promoters for CO2-hydrate formation: Thermodynamics properties, and kinetics in presence of sodium dodecyl sulfate.
Torre JP, et al.
Chemical Engineering Science, 126, 688-697 (2015)
Practical Synthesis of Di-tert-Butyl-Phosphinoferrocene.
Busacca CA, et al.
Organic Syntheses, 90, 316-326 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service