All Photos(2)
About This Item
Linear Formula:
C2H5OCH2CH2CO2C2H5
CAS Number:
Molecular Weight:
146.18
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
166 °C (lit.)
Recommended Products
vapor density
5.03 (vs air)
Quality Level
Assay
99%
form
liquid
impurities
≤0.05% (water)
refractive index
n20/D 1.405 (lit.)
bp
166 °C (lit.)
mp
-75 °C
solubility
water: soluble 54.1 g/L at 20 °C
density
0.95 g/mL at 25 °C (lit.)
SMILES string
CCOCCC(=O)OCC
InChI
1S/C7H14O3/c1-3-9-6-5-7(8)10-4-2/h3-6H2,1-2H3
InChI key
BHXIWUJLHYHGSJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Used as solvent in preparing new polymers.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
136.4 °F
Flash Point(C)
58 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of Applied Polymer Science. Applied Polymer Symposium, 88, 636-640 (2003)
J. Photopolym. Sci. Technol., 16, 335-345 (2003)
B Qi et al.
Applied microbiology and biotechnology, 58(5), 684-689 (2002-04-17)
Five fungal species, Cladosporium resinae (ATCC 34066), Cladosporium sphaerospermum (ATCC 200384), Exophiala lecanii-corni (CBS 102400), Mucor rouxii (ATCC 44260), and Phanerochaete chrysosporium (ATCC 24725), were tested for their ability to degrade nine compounds commonly found in industrial off-gas emissions. Fungal
The hydroxyl radical reaction rate constant and products of ethyl 3-ethoxypropionate.
Baxley JS, et al.
International Journal of Chemical Kinetics, 29(8), 637-644 (1997)
P J Deisinger et al.
Xenobiotica; the fate of foreign compounds in biological systems, 20(10), 989-997 (1990-10-01)
1. The metabolism and disposition of ethyl 3-ethoxypropionate (EEP) in male Sprague-Dawley rats was studied following single oral gavage at 150 or 1500 mg/kg. The 14C-EEP was rapidly absorbed at both dose levels, and was excreted predominantly as metabolites in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service