Skip to Content
Merck
All Photos(1)

Key Documents

B103608

Sigma-Aldrich

γ-Butyrolactone

ReagentPlus®, ≥99%

Synonym(s):

gamma-Butyrolactone, γ-Hydroxybutyric acid lactone, 4-Hydroxybutyric acid lactone, GBL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09
Beilstein:
105248
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3 (vs air)

Quality Level

vapor pressure

1.5 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

851 °F

expl. lim.

16 %

refractive index

n20/D 1.436 (lit.)

bp

204-205 °C (lit.)

mp

−45 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCO1

InChI

1S/C4H6O2/c5-4-2-1-3-6-4/h1-3H2

InChI key

YEJRWHAVMIAJKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For the introduction of 3-carboxypropyl side chain. Used as a component of electrolyte solutions in batteries and capacitors.

Biochem/physiol Actions

Precursor of γ-hydroxybutyric acid (GHB). It blocks dopamine release by blocking impulse flow in dopaminergic neurons. Pretreatment with γ-butyrolactone allows detection of autoreceptor-induced dopamine release.
Precursor of γ-hydroxybutyric acid (GHB); blocks dopamine release.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Power Sources, 43, 195-195 (1993)
S Ohkawa et al.
Journal of medicinal chemistry, 34(1), 267-276 (1991-01-01)
A novel series of (3-pyridylmethyl)benzoquinone derivatives was molecular designed and synthesized for the dual purpose of inhibiting thromboxane A2 and leukotriene biosynthesis enzymes and scavenging active oxygen species (AOS). They were evaluated for inhibition of TXA2 synthase, inhibition of 5-lipoxygenase
Monica Blichowski et al.
Epilepsia, 56(7), 1081-1087 (2015-06-03)
Infantile spasms (or IS) is a catastrophic childhood epilepsy that is particularly prevalent in children with Down syndrome. Previously, we have shown that the Ts65Dn (Ts) mouse model of Down syndrome is a useful substrate upon which to develop an
Bo Xue et al.
Biochemistry, 52(13), 2359-2370 (2013-03-07)
The in vitro evolution and engineering of quorum-quenching lactonases with enhanced reactivities was achieved using a thermostable GKL enzyme as a template, yielding the E101G/R230C GKL mutant with increased catalytic activity and a broadened substrate range [Chow, J. Y., Xue
Zheng-Yuan Su et al.
Chemical research in toxicology, 26(3), 477-485 (2013-02-28)
Cancer development has been linked to epigenetic modifications of cancer oncogenes and tumor suppressor genes; in advanced metastatic cancers, severe epigenetic modifications are present. We previously demonstrated that the progression of prostate tumors in TRAMP mice is associated with methylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service