Skip to Content
Merck
All Photos(3)

Key Documents

431346

Sigma-Aldrich

Ammonium molybdate tetrahydrate

ACS reagent, 99.98% trace metals basis

Synonym(s):

Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6Mo7O24 · 4H2O
CAS Number:
Molecular Weight:
1235.86
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.55

grade

ACS reagent

Quality Level

Agency

suitable for SM 5210

description

for inorganic trace analysis

Assay

81.0-83.0% MoO3 basis
99.98% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: molybdenum

impurities

≤0.005% insolubles

density

2.498 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
nitrate (NO3-): passes test
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.02%

cation traces

Mg and allied cations: ≤0.01%
heavy metals: ≤0.001% (as lead)

SMILES string

N.N.N.N.N.N.O.O.O.O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O

InChI

1S/7Mo.6H3N.10H2O.18O/h;;;;;;;6*1H3;10*1H2;;;;;;;;;;;;;;;;;;/q;;;;3*+2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-6

InChI key

FIXLYHHVMHXSCP-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium molybdate tetrahydrate (AMT) is a molybdenum salt. Study of its extraction from nickel-molybdenum (Ni–Mo) ore by direct hydrometallurgical method has been reported. Its degradation by combustion to form elemental molybdenum powder using metallic reducers has been investigated.

Application

Ammonium molybdate tetrahydrate may be used in the following processes:
  • As a precursor in the generation of calcium molybdate (CaMoO4) spherulites.
  • Synthesis of porous carbon-supported molybdenum dioxide (MoO2).
  • Preparation of Iodine-iodide-molybdate solution to determine glucose level by glucose oxidase method.
  • Preparation of polymer bulk heterojunction solar cells.
  • As a precursor in the synthesis of Mo-N-codoped TiO2 (titanium dioxide).
  • Synthesis of MoBi2S5 (molybdenum bismuth sulphide) thin films.
Applied in the ultrasonic synthesis of phase-pure praseodymium molybdate, Pr2Mo3O12.
Reagent used for oxidation of sulfides to sulfonyls.

Features and Benefits

Meets A.C.S. reagent specificatons.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-situ preparation of porous carbon-supported molybdenum dioxide and its performance in the oxidative desulfurization of thiophene.
Zhuang L, et al.
J. Mater. Sci., 49(16), 5606-5616 (2014)
Organic photovoltaic devices incorporating a molybdenum oxide hole-extraction layer deposited by spray-coating from an ammonium molybdate tetrahydrate precursor.
Griffin J, et al.
Organic Electronics, 15(3), 692-700 (2014)
Enhanced visible light activity and mechanism of TiO2 codoped with molybdenum and nitrogen.
Cheng X, et al.
Materials Science and Engineering, B, 178(7), 425-430 (2013)
Tetrahedron, 62, 11592-11592 (2006)
Recovery and purification of molybdenum from Ni-Mo ore by direct air oxidation in alkaline solution.
Zhao Z, et al.
Hydrometallurgy, 103(1), 68-73 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service