Skip to Content
Merck
All Photos(2)

Key Documents

31277

Sigma-Aldrich

Cobalt(II) chloride hexahydrate

puriss. p.a., ACS reagent, reag. Ph. Eur., 98-102%

Synonym(s):

Cobaltous chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCl2 · 6H2O
CAS Number:
Molecular Weight:
237.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
form:
solid

grade

ACS reagent
puriss. p.a.

Agency

reag. Ph. Eur.

vapor pressure

40 mmHg ( 0 °C)

Assay

98-102%

form

solid

impurities

≤0.01% water-insoluble matter

anion traces

nitrate (NO3-): ≤100 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cu: ≤20 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤50 mg/kg
Na: ≤100 mg/kg
Ni: ≤500 mg/kg
Pb: ≤5 mg/kg
Zn: ≤25 mg/kg

SMILES string

O.O.O.O.O.O.Cl[Co]Cl

InChI

1S/2ClH.Co.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

GFHNAMRJFCEERV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cobalt(II) chloride hexahydrate is a hydrated form of cobalt chloride. It is used as a catalyst for various organic reactions such as acetylation and tosylation of alcohols, chemoselective reduction of carboxylic esters, and condensation reactions.

Application

Cobalt(II) chloride hexahydrate can be used as a catalyst:      
  • To convert various aldoximes into corresponding nitriles in the presence of an inorganic base.      
  • To prepare organic-inorganic polyvinylidene fluoride nanofiber composites (PVDF/CoCl2) for the production of pure hydrogen production from NaBH4.

It can also be used as a cobalt precursor to synthesize nickel-cobalt-based binary metal phosphide (NiCoP) hollow spheres, which are used for electrocatalysis and energy storage (supercapacitor) applications.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gláucia Melo Reis et al.
Pharmacology, biochemistry, and behavior, 131, 112-118 (2015-02-18)
The stimulation of the retrosplenial cortex (RSC) is antinociceptive in the rat tail-flick and formalin tests. The rat RSC is caudal to and send projections to the ipsilateral anterior cingulate cortex (ACC), which is also involved in pain processing. This
H Lawrence et al.
The bone & joint journal, 96-B(9), 1172-1177 (2014-09-04)
Abnormal wear of cobalt-containing metal-on-metal joints is associated with inflammatory pseudotumours. Cobalt ions activate human toll-like receptor 4 (TLR4), which normally responds to bacterial lipopolysaccharide (LPS) in sepsis. Activation of TLR4 by LPS increases the expression of chemokines IL-8 and
Junya Nagai et al.
Biochemical and biophysical research communications, 450(1), 476-481 (2014-06-14)
Recently, we found that albumin overload induces expression of the transcription factor hypoxia-inducible factor-1α (HIF-1α) protein and several HIF-1 target genes in human renal proximal tubular epithelial cell line HK-2. In this study, the role of albumin-bound fatty acids in
Senthilguru Kulanthaivel et al.
Materials science & engineering. C, Materials for biological applications, 58, 648-658 (2015-10-20)
The present study delineates the synthesis and characterization of cobalt doped proangiogenic-osteogenic hydroxyapatite. Hydroxyapatite samples, doped with varying concentrations of bivalent cobalt (Co(2+)) were prepared by the ammoniacal precipitation method and the extent of doping was measured by ICP-OES. The
Vaidehi Agrawal et al.
Melanoma research, 24(6), 556-567 (2014-10-12)
Metastatic melanoma is a deadly form of cancer with few therapeutic options and the cause of more than 9480 deaths annually in the USA alone. Novel treatment options for this disease are urgently needed. Here we test the efficacy of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service