Skip to Content
Merck
All Photos(1)

Documents

30402

Sigma-Aldrich

Ammonium phosphate dibasic

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)

Synonym(s):

Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (sec)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2HPO4
CAS Number:
Molecular Weight:
132.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent
puriss. p.a.

Agency

reag. Ph. Eur.

Assay

≥98% (alkalimetric)

form

powder, crystals or granules

impurities

≤0.001% heavy metals (as Pb)

pH

7.7-8.1 (25 °C, 5%)

mp

155 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.003%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.001%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.005%

storage temp.

room temp

SMILES string

N.N.OP(O)(O)=O

InChI

1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)

InChI key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Human Bone Paleoproteomics Utilizing the Single-Pot, Solid-Phase-Enhanced Sample Preparation Method to Maximize Detected Proteins and Reduce Humics.: This study demonstrates the application of ammonium phosphate dibasic in the preparation of human bone samples for paleoproteomics. The method enhances protein detection and reduces contamination, significantly improving the analysis of ancient proteins (Cleland, 2018).

  • Effect of Mg(2+) doping on beta-alpha phase transition in tricalcium phosphate (TCP) bioceramics.: Ammonium phosphate dibasic is utilized in the synthesis of Mg-doped tricalcium phosphate bioceramics. The study investigates the phase transition behavior, which is critical for developing advanced biomaterials for medical applications (Frasnelli & Sglavo, 2016).

  • Development and validation of a stability-indicating HPLC method for determination of voriconazole and its related substances.: The research outlines the use of ammonium phosphate dibasic in the development of a stability-indicating HPLC method for analyzing voriconazole. This method ensures accurate detection of the drug and its impurities, crucial for pharmaceutical quality control (Gu & Li, 2009).

  • Crystallization and preliminary crystallographic analysis of the catalytic domain cellobiohydrolase I from Talaromyces emersonii.: Ammonium phosphate dibasic is employed in the crystallization process of cellobiohydrolase I. The study provides insights into the enzyme′s structure, aiding in the understanding of its catalytic mechanisms and potential industrial applications (Grassick et al., 2003).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Sakakura et al.
Journal of chromatography, 621(2), 123-131 (1993-11-24)
A method for the simultaneous determination of bile acids in rat bile and serum by high-performance liquid chromatography with a post-column enzymic reaction and fluorescence detection has been developed. Without prior fractionation and alkaline hydrolysis, 26 unconjugated, glycine- and taurine-conjugated
Electric Field-Assisted Solid Phase Extraction: Study of Electrochromatographic Parameters with an Anionic Model Compound.
Orlando RM, et al.
Journal of the Brazilian Chemical Society, 26(2), 310-318 (2015)
Hydrothermal synthesis, structures and luminescent properties of nanocrystaline Ca8Gd2(PO4)6O2: Eu2+, Eu3+ phosphors.
Bharat L, et al.
Chemical Engineering Journal, 240, 179-186 (2014)
Everest Akaraonye et al.
Biotechnology journal, 7(2), 293-303 (2011-12-08)
The main hindrance in the use of polyhydroxyalkanoates (PHAs) as a replacement for existing petroleum-based plastics is their high production cost. The carbon source accounts for 50% of the cost for PHA production. Thus, increasing the yield and productivity of
G A Morán-Marroquín et al.
International journal of food microbiology, 151(1), 87-92 (2011-09-10)
Knowledge of physiological behavior of indigenous tequila yeast used in fermentation process is still limited. Yeasts have significant impact on the productivity fermentation process as well as the sensorial characteristics of the alcoholic beverage. For these reasons a better knowledge

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service