Skip to Content
Merck
All Photos(1)

Key Documents

24587

Sigma-Aldrich

Petroleum ether

puriss., low boiling point hydrogen treated naphtha, meets analytical specification of DAB, bpmin. 75% 40-60 °C (min. 75%)

Synonym(s):

Petroleum benzin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.07
bp:
40-60 °C (min. 75%)
vapor pressure:
25.8 psi

vapor density

2.5 (vs air)

Quality Level

vapor pressure

25.8 psi

grade

puriss.

form

liquid

quality

meets analytical specification of DAB

impurities

≤0.001% benzene (GC)
≤0.001% non-volatile matter
≤0.005% S-compounds (as S)
≤0.01% water (Karl Fischer)
≤2% n-hexane (GC)

bp

40-60 °C (min. 75%)

density

0.642-0.656 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

Application


  • Bioassay-Guided Skin-Beneficial Effects of Fractionated Sideritis raeseri subsp. raeseri Extract.: This study explores the skin-beneficial effects of Sideritis raeseri, using petroleum ether for the fractionation of active components, showing potential in dermatological applications (Krgović et al., 2022).

  • Physico-Chemical Analysis and Fatty Acid Profiling of Fenugreek (Trigonella foenum-graecum) Seed Oil Using Different Solvents.: This research assesses the impact of various solvents including petroleum ether on the extraction and fatty acid profile of fenugreek seed oil, emphasizing analytical techniques in chemical analysis (Munshi et al., 2020).

  • Poplar bark lipids enhance mouse immunity by inducing T cell proliferation and differentiation.: The study investigates the immunomodulatory effects of lipids extracted from poplar bark using petroleum ether, underscoring its utility in biochemical extractions (Tang et al., 2020).

  • Phytochemical screening, cytotoxic and antimicrobial activities of Limonium socotranum and Peperomia blanda extracts.: This article details the use of petroleum ether in the extraction process for studying the cytotoxic and antimicrobial properties of Limonium socotranum and Peperomia blanda (Al-Madhagi et al., 2019).

  • The study of metabolites from fermentation culture of Alternaria oxytropis.: Focuses on identifying metabolites in the fermentation culture of Alternaria oxytropis, with petroleum ether used to extract lipid-soluble compounds, enhancing understanding of microbial metabolism in an analytical context (Song et al., 2019).

Other Notes

The article number 24587-4X2.5L-R will be discontinued. Please order the single bottle 24587-2.5L-R which is physically identical with the same exact specifications.
The article number 24587-4X5L-R will be discontinued. Please order the single bottle 24587-5L-R which is physically identical with the same exact specifications.
The article number 24587-6X1L-M will be discontinued. Please order the single bottle 24587-1L-M which is physically identical with the same exact specifications.
The article number 24587-6X1L-R will be discontinued. Please order the single bottle 24587-1L-R which is physically identical with the same exact specifications.
Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<-40.0 °F - closed cup

Flash Point(C)

< -40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S E Besra et al.
Journal of ethnopharmacology, 54(1), 1-6 (1996-10-01)
Pharmacological studies were conducted with the petroleum ether extract of leaves of the plant Litchi chinensis Gaertn. (Sapindaceae) on experimental animals. The extract was found to possess antiinflammatory, analgesic and antipyretic activity. Acute toxicity studies revealed that the extract, up
Rapid chromatographic technique for preparative separations with moderate resolution.
Still WC, et al.
The Journal of Organic Chemistry, 43(14), 2923-2925 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service