Skip to Content
Merck
All Photos(1)

Key Documents

32299

Sigma-Aldrich

Petroleum ether

puriss. p.a., ACS reagent, reag. ISO, low boiling point hydrogen treated naphtha, bp≥ 90% 40-60 °C (≥ 90%)

Synonym(s):

Hydrotreated light naphtha

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.21
bp:
40-60 °C (≥ 90%)
vapor pressure:
25.8 psi

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. ISO

vapor density

2.5 (vs air)

vapor pressure

25.8 psi

form

liquid

impurities

≤0.001% non-volatile matter
≤0.002% free acid (as CH3COOH)
≤0.005% S-compounds (as S)
≤0.01% benzene (GC)
≤0.01% water (Karl Fischer)

bp

40-60 °C (≥ 90%)

iodine value

≤0.3

density

0.640-0.655 g/mL at 20 °C

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.02 mg/kg
Sn: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

Looking for similar products? Visit Product Comparison Guide

General description

Petroleum ether is a low boiling point fraction having boiling point in the range of 40-60°C. Extracts of leaves of Litchi chinensis Gaertn. (Sapindaceae) have been prepared in petroleum ether and were evaluated for antiinflammatory, analgesic and antipyretic activities.

Application

Petroleum ether may be used in absorption chromatographic purification of organic compounds.

Other Notes

The article number 32299-4X2.5L will be discontinued. Please order the single bottle 32299-2.5L which is physically identical with the same exact specifications.
The article number 32299-4X2.5L-M will be discontinued. Please order the single bottle 32299-2.5L-M which is physically identical with the same exact specifications.
The article number 32299-6X1L will be discontinued. Please order the single bottle 32299-1L which is physically identical with the same exact specifications.
The article number 32299-6X1L-M will be discontinued. Please order the single bottle 32299-1L-M which is physically identical with the same exact specifications.
Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

<-5.8 °F

Flash Point(C)

< -21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid chromatographic technique for preparative separations with moderate resolution.
Still WC, et al.
The Journal of Organic Chemistry, 43(14), 2923-2925 (1978)
S E Besra et al.
Journal of ethnopharmacology, 54(1), 1-6 (1996-10-01)
Pharmacological studies were conducted with the petroleum ether extract of leaves of the plant Litchi chinensis Gaertn. (Sapindaceae) on experimental animals. The extract was found to possess antiinflammatory, analgesic and antipyretic activity. Acute toxicity studies revealed that the extract, up

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service