Skip to Content
Merck
All Photos(3)

Key Documents

238511

Sigma-Aldrich

Potassium permanganate

≤150 μm particle size, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
EC Number:
MDL number:
UNSPSC Code:
12352312
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

97%

form

powder, crystals or chunks

reaction suitability

reagent type: oxidant

particle size

≤150 μm

pH

7.2-9.7 (20 °C, 20 g/L)

SMILES string

[K+].[O-][Mn](=O)(=O)=O

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Layered MnO2 with very promising characteristics for battery applications is produced by thermal decomposition of KMnO4.
Suitable for heterogeneous oxidations.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kim, S.H. et al.
Chemistry of Materials, 11, 557-557 (1999)
Crown polyether chemistry. Potassium permanganate oxidations in benzene.
Sam DJ and Simmons HE.
Journal of the American Chemical Society, 94(11), 4024-4025 (1972)
Eagleson M.
Concise Encyclopedia Chemistry, 227-227 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 1047-1047 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 622-622 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service