Skip to Content
Merck
All Photos(4)

Key Documents

232653

Sigma-Aldrich

Chromium(VI) oxide

ReagentPlus®, 99.9% trace metals basis

Synonym(s):

Chromic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CrO3
CAS Number:
Molecular Weight:
99.99
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.55
Assay:
99.9% trace metals basis
form:
(powder and or flakes)

Quality Level

product line

ReagentPlus®

Assay

99.9% trace metals basis

form

(powder and or flakes)

pH

<1 (20 °C, 100 g/L)

mp

196 °C (dec.) (lit.)

SMILES string

O=[Cr](=O)=O

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Chromium (VI) oxide is acidic anhydride of chromic acid. It is a useful oxidizing reagent for the conversions of:,Carbon-hydrogen bonds to alcohols,,Alkenes to α, β-unsaturated ketones,Alcohol to aldehydes, acids, and keto acids.

Application

Chromium (VI) oxide may be employed as a catalyst for the following reactions:
  • Transformation of various primary alcohols to carboxylic acids.
  • Oxidation of arenes (naphthalenes and anthrathene) to the corresponding quinones by using periodic acid as the terminal oxidant in acetonitrile.
  • Benzylic oxidation of various substituted toluene to the corresponding benzoic acids.
  • Oxidation of sulfides to sulfones.

Used with ammonium chloride to oxidize hydrobenzoins to benzils.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Letters in Organic Chemistry, 3, 842-842 (2006)
Chromium (VI) Oxide.
Freeman F, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2008)
Chromium (VI) oxide-catalyzed oxidation of arenes with periodic acid.
Yamazaki S.
Tetrahedron Letters, 42(19), 3355-3357 (2001)
Reem H Elbekai et al.
Toxicology in vitro : an international journal published in association with BIBRA, 22(5), 1184-1190 (2008-05-14)
NAD(P)H:quinone oxidoreductase (Nqo1)-mediated detoxification of quinones plays a critical role in cancer prevention. Metals alter the carcinogenicity of AhR ligands, such as TCDD, by modulating the induction of Nqo1, but the mechanism(s) remain unresolved. To decipher the molecular mechanisms involved
Reem H Elbekai et al.
Free radical biology & medicine, 39(11), 1499-1511 (2005-11-09)
Heavy metals alter the capacity of AhR ligands to induce the bioactivating phase I and the detoxifying phase II xenobiotic-metabolizing enzymes but the mechanism(s) remain unknown. In the present study, we evaluated the role of As(3+)-, Cd(2+)-, and Cr(6+)-induced oxidative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service