Skip to Content
Merck
All Photos(3)

Documents

221716

Sigma-Aldrich

Ammonium oxalate monohydrate

ACS reagent, ≥99%

Synonym(s):

Oxalic acid diammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2C2O4 · H2O
CAS Number:
Molecular Weight:
142.11
Beilstein:
3759830
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

vapor pressure

<0.1 hPa ( 20 °C)

Assay

≥99%
99.0-101.0% (ACS specification)

form

crystalline powder
crystals or granules

impurities

≤0.005% insolubles

ign. residue

≤0.02%

pH

6-7 (25 °C, 14.2 g/L)

mp

133 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤2 ppm
heavy metals: ≤5 ppm (by ICP)

SMILES string

OC(C(O)=O)=O.N.O.N

InChI

1S/C2H2O4.2H3N.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);2*1H3;1H2

InChI key

MSMNVXKYCPHLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium oxalate monohydrate is a diammonium salt of oxalic acid. Its refined crystal structure has been investigated by three-dimensional diffraction studies. Influence of bivalent [Mn(II), Co(II), Ni(II)] and trivalent cations [Fe(III), Cr(III)] impurities on the growth rates of ammonium oxalate monohydrate crystals has been described. Its space group and unit cell parameters have been analyzed by neutron diffraction studies. Its IR spectra have been compared with that of isolated coal-solubilizing agent (CSA).

Application

Ammonium oxalate monohydrate may be used in the preparation of tetramethylammonium oxalate.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeonho Kim et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(46), 10510-10518 (2020-03-27)
In this work, environmentally friendly photocatalysts with attractive catalytic properties are reported that have been prepared by introducing SnO2 quantum dots (QDs) directly onto ZnSe(N2 H4 )0.5 substrates to induce advantageous charge separation. The SnO2 /ZnSe(N2 H4 )0.5 nanocomposites could
A convenient method for preparation of quaternary ammonium salts.
Markowitz M.
The Journal of Organic Chemistry, 22(8), 983-984 (1957)
Sina Matavos-Aramyan et al.
Free radical biology & medicine, 106, 158-167 (2017-02-19)
Three novel modified advanced oxidation process systems including ascorbic acid-, pro-oxidants- and ascorbic acid-pro-oxidants-modified Fenton system were utilized to study the disinfection efficiency on Cryptosporidium-contaminated drinking water samples. Different concentrations of divalent and trivalent iron ions, hydrogen peroxide, ascorbic acid
M S Cohen et al.
Applied and environmental microbiology, 56(11), 3285-3291 (1990-11-01)
Low-ranked coals were dissolved by using cell extracts derived from liquid cultures of Trametes versicolor. The coal-solubilizing agent (CSA) was separated from the broth components by a multistep isolation procedure including reverse-phase high-pressure liquid chromatography, size exclusion chromatography, ethanol fractionation
Neutron diffraction study of ammonium oxalate monohydrate,(NH4)2C2O4.H2O.
Padmanabhan VM, et al.
Acta Crystallographica, 18(3), 567-568 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service