Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium oxalate monohydrate
Synonym(s):
Oxalic acid diammonium salt
Linear Formula:
(NH
4
)
2
C
2
O
4
· H
2
O
CAS Number:
6009-70-7
Molecular Weight:
142.11
EC Number:
238-135-4
Products
Product Category
salts (2)
building blocks (1)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (1)
Puriss P.A. (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
221716
Ammonium oxalate monohydrate,
ACS reagent, ≥99%
Expand
View Pricing
32304
Ammonium oxalate monohydrate,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-101.0%
Expand
View Pricing
379743
Ammonium oxalate monohydrate,
≥99.99% trace metals basis
Expand
View Pricing
09898
Ammonium oxalate monohydrate,
BioUltra, ≥99.5% (RT)
Expand
View Pricing